Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 103344863-103345120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCGF6
Entrez Gene ID
84108
Gene Name
polycomb group ring finger 6
Gene Aliases
MBLR, RNF134
UniGene
Hs.335808
Gene Chromosome Location
Chr.10: 103302796-103351134 on Build GRCh38
Transcripts
NM_001011663.1 NM_032154.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5177983 c.782+6T>C p.? Chr.10: 103345018-103345018 on Build GRCh38
3433842 c.736C>T p.R246C Chr.10: 103345070-103345070 on Build GRCh38
6874620 c.737G>A p.R246H Chr.10: 103345069-103345069 on Build GRCh38
343389 c.761C>T p.S254F Chr.10: 103345045-103345045 on Build GRCh38
4912083 c.770T>C p.L257P Chr.10: 103345036-103345036 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745373135 Chr.10: 103345001-103345001 on Build GRCh38
rs889603867 Chr.10: 103345032-103345032 on Build GRCh38
rs1051810552 Chr.10: 103344914-103344914 on Build GRCh38
rs770464250 Chr.10: 103344973-103344973 on Build GRCh38
rs764115744 Chr.10: 103345026-103345026 on Build GRCh38
rs756417949 Chr.10: 103345055-103345055 on Build GRCh38
rs778127531 Chr.10: 103345086-103345086 on Build GRCh38
rs904437976 Chr.10: 103344957-103344959 on Build GRCh38
rs769240846 Chr.10: 103345002-103345002 on Build GRCh38
rs372984226 Chr.10: 103345050-103345050 on Build GRCh38
rs61760880 Chr.10: 103345062-103345062 on Build GRCh38
rs149394518 Chr.10: 103345060-103345060 on Build GRCh38
rs372664700 Chr.10: 103345096-103345096 on Build GRCh38
rs542998177 Chr.10: 103344947-103344947 on Build GRCh38
rs774988597 Chr.10: 103345005-103345005 on Build GRCh38
rs773997289 Chr.10: 103345027-103345027 on Build GRCh38
rs137910892 Chr.10: 103345069-103345069 on Build GRCh38
rs758860265 Chr.10: 103345076-103345076 on Build GRCh38
rs998105864 Chr.10: 103345016-103345016 on Build GRCh38
rs767629086 Chr.10: 103345049-103345049 on Build GRCh38
rs762840265 Chr.10: 103345014-103345014 on Build GRCh38
rs1044154390 Chr.10: 103344910-103344910 on Build GRCh38
rs138884621 Chr.10: 103345028-103345028 on Build GRCh38
rs369977135 Chr.10: 103345058-103345058 on Build GRCh38
rs904201596 Chr.10: 103344924-103344924 on Build GRCh38
rs962689338 Chr.10: 103345082-103345082 on Build GRCh38
rs754001394 Chr.10: 103345059-103345059 on Build GRCh38
rs555197349 Chr.10: 103344986-103344986 on Build GRCh38
rs1042907992 Chr.10: 103344950-103344950 on Build GRCh38
rs776383200 Chr.10: 103344978-103344978 on Build GRCh38
rs769470053 Chr.10: 103345098-103345098 on Build GRCh38
rs182979232 Chr.10: 103344928-103344928 on Build GRCh38
rs35956583 Chr.10: 103345094-103345094 on Build GRCh38
rs935639228 Chr.10: 103344926-103344926 on Build GRCh38
rs933377326 Chr.10: 103344895-103344895 on Build GRCh38
rs891673432 Chr.10: 103345013-103345013 on Build GRCh38

View Full Product Details