Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18977686-18977933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83G
Entrez Gene ID
644815
Gene Name
family with sequence similarity 83 member G
Gene Aliases
PAWS1
UniGene
Hs.710727
Gene Chromosome Location
Chr.17: 18968789-19006144 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001039999.2 XM_017024953.1 NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587781 c.1851G>A p.E617E Chr.17: 18977815-18977815 on Build GRCh38
1609930 c.1895G>A p.R632H Chr.17: 18977771-18977771 on Build GRCh38
6660243 c.1917C>T p.N639N Chr.17: 18977749-18977749 on Build GRCh38
295077 c.1850_1851insT p.E617fs*93 Chr.17: 18977815-18977816 on Build GRCh38
5620790 c.1883C>T p.P628L Chr.17: 18977783-18977783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753077917 Chr.17: 18977846-18977847 on Build GRCh38
rs752126627 Chr.17: 18977731-18977731 on Build GRCh38
rs371125554 Chr.17: 18977749-18977749 on Build GRCh38
rs1052398432 Chr.17: 18977785-18977785 on Build GRCh38
rs758772623 Chr.17: 18977729-18977729 on Build GRCh38
rs766692916 Chr.17: 18977741-18977741 on Build GRCh38
rs896136965 Chr.17: 18977851-18977851 on Build GRCh38
rs771017043 Chr.17: 18977866-18977866 on Build GRCh38
rs776514603 Chr.17: 18977876-18977876 on Build GRCh38
rs376311708 Chr.17: 18977899-18977899 on Build GRCh38
rs752034805 Chr.17: 18977805-18977805 on Build GRCh38
rs752937192 Chr.17: 18977782-18977782 on Build GRCh38
rs773273233 Chr.17: 18977764-18977764 on Build GRCh38
rs775766698 Chr.17: 18977896-18977896 on Build GRCh38
rs764496911 Chr.17: 18977794-18977794 on Build GRCh38
rs572824194 Chr.17: 18977748-18977748 on Build GRCh38
rs905300005 Chr.17: 18977836-18977836 on Build GRCh38
rs779496696 Chr.17: 18977806-18977806 on Build GRCh38
rs1004418613 Chr.17: 18977884-18977884 on Build GRCh38
rs34552844 Chr.17: 18977745-18977746 on Build GRCh38
rs368202632 Chr.17: 18977900-18977900 on Build GRCh38
rs201668455 Chr.17: 18977727-18977727 on Build GRCh38
rs1014037928 Chr.17: 18977796-18977796 on Build GRCh38
rs751950262 Chr.17: 18977904-18977904 on Build GRCh38
rs372021645 Chr.17: 18977846-18977846 on Build GRCh38
rs1056183148 Chr.17: 18977847-18977848 on Build GRCh38
rs772063783 Chr.17: 18977754-18977754 on Build GRCh38
rs759663540 Chr.17: 18977883-18977883 on Build GRCh38
rs759629345 Chr.17: 18977777-18977777 on Build GRCh38
rs763298260 Chr.17: 18977784-18977784 on Build GRCh38
rs201172910 Chr.17: 18977722-18977722 on Build GRCh38
rs924496501 Chr.17: 18977779-18977779 on Build GRCh38
rs200038747 Chr.17: 18977726-18977726 on Build GRCh38
rs965059488 Chr.17: 18977888-18977888 on Build GRCh38
rs367677167 Chr.17: 18977736-18977736 on Build GRCh38
rs769831705 Chr.17: 18977885-18977885 on Build GRCh38
rs771103367 Chr.17: 18977772-18977772 on Build GRCh38
rs577449943 Chr.17: 18977861-18977861 on Build GRCh38
rs374490985 Chr.17: 18977771-18977771 on Build GRCh38
rs746904439 Chr.17: 18977865-18977865 on Build GRCh38
rs1027117583 Chr.17: 18977847-18977847 on Build GRCh38
rs772271859 Chr.17: 18977886-18977886 on Build GRCh38
rs373827776 Chr.17: 18977732-18977732 on Build GRCh38
rs368737975 Chr.17: 18977908-18977908 on Build GRCh38
rs201669881 Chr.17: 18977730-18977730 on Build GRCh38
rs773363329 Chr.17: 18977714-18977714 on Build GRCh38
rs201046878 Chr.17: 18977778-18977778 on Build GRCh38
rs754522335 Chr.17: 18977843-18977843 on Build GRCh38
rs972543410 Chr.17: 18977770-18977770 on Build GRCh38
rs753405420 Chr.17: 18977824-18977824 on Build GRCh38
rs766625194 Chr.17: 18977719-18977719 on Build GRCh38
rs376662345 Chr.17: 18977905-18977905 on Build GRCh38
rs771971752 Chr.17: 18977858-18977858 on Build GRCh38
rs747982022 Chr.17: 18977753-18977753 on Build GRCh38
rs747816872 Chr.17: 18977849-18977849 on Build GRCh38

View Full Product Details