Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93685081-93685318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
UniGene
Hs.586650
Gene Chromosome Location
Chr.10: 93667883-93702572 on Build GRCh38
Transcripts
NM_145246.4 XM_005269515.4 XM_005269516.4 XM_011539259.2 XM_011539260.2 XM_017015656.1 XM_017015657.1 XM_017015658.1 XM_017015659.1 XM_017015660.1 XM_017015661.1 XM_017015662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675445 c.614T>G p.L205R Chr.10: 93685257-93685257 on Build GRCh38
4786470 c.580G>A p.G194S Chr.10: 93685291-93685291 on Build GRCh38
1675444 c.614T>G p.L205R Chr.10: 93685257-93685257 on Build GRCh38
1349882 c.611C>T p.A204V Chr.10: 93685260-93685260 on Build GRCh38
1638694 c.603G>T p.K201N Chr.10: 93685268-93685268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750462341 Chr.10: 93685250-93685250 on Build GRCh38
rs890024265 Chr.10: 93685117-93685117 on Build GRCh38
rs761981204 Chr.10: 93685275-93685275 on Build GRCh38
rs577644013 Chr.10: 93685163-93685163 on Build GRCh38
rs1030807252 Chr.10: 93685165-93685165 on Build GRCh38
rs150512803 Chr.10: 93685223-93685223 on Build GRCh38
rs772775452 Chr.10: 93685229-93685229 on Build GRCh38
rs372905927 Chr.10: 93685222-93685222 on Build GRCh38
rs776731975 Chr.10: 93685268-93685268 on Build GRCh38
rs79118451 Chr.10: 93685148-93685148 on Build GRCh38
rs201575957 Chr.10: 93685232-93685232 on Build GRCh38
rs199662472 Chr.10: 93685217-93685217 on Build GRCh38
rs767633444 Chr.10: 93685246-93685250 on Build GRCh38
rs1006859758 Chr.10: 93685125-93685126 on Build GRCh38
rs779951055 Chr.10: 93685202-93685202 on Build GRCh38
rs768641121 Chr.10: 93685264-93685264 on Build GRCh38
rs773588352 Chr.10: 93685214-93685217 on Build GRCh38
rs765474094 Chr.10: 93685278-93685278 on Build GRCh38
rs200966941 Chr.10: 93685213-93685213 on Build GRCh38
rs767701 Chr.10: 93685174-93685174 on Build GRCh38
rs1043338689 Chr.10: 93685115-93685115 on Build GRCh38
rs751134115 Chr.10: 93685207-93685207 on Build GRCh38
rs181127998 Chr.10: 93685135-93685135 on Build GRCh38
rs899855940 Chr.10: 93685108-93685108 on Build GRCh38
rs773047436 Chr.10: 93685288-93685288 on Build GRCh38
rs772237500 Chr.10: 93685197-93685197 on Build GRCh38
rs149181398 Chr.10: 93685279-93685279 on Build GRCh38
rs904606739 Chr.10: 93685151-93685151 on Build GRCh38
rs546505041 Chr.10: 93685273-93685273 on Build GRCh38
rs746223745 Chr.10: 93685227-93685227 on Build GRCh38
rs747462128 Chr.10: 93685251-93685251 on Build GRCh38

View Full Product Details