Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 41192543-41192816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD7
Entrez Gene ID
84078
Gene Name
kelch repeat and BTB domain containing 7
Gene Aliases
-
UniGene
Hs.63841
Gene Chromosome Location
Chr.13: 41191575-41194566 on Build GRCh38
Transcripts
NR_120423.1

Gene Symbol
LOC101929140
Entrez Gene ID
101929140
Gene Name
uncharacterized LOC101929140
Gene Aliases
-
UniGene
Hs.663088
Gene Chromosome Location
Chr.13: 41132928-41236686 on Build GRCh38
Transcripts
NR_120423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690377 c.1598T>G p.V533G Chr.13: 41192660-41192660 on Build GRCh38
5714589 c.1570G>A p.E524K Chr.13: 41192688-41192688 on Build GRCh38
3987400 c.1446T>A p.V482V Chr.13: 41192812-41192812 on Build GRCh38
3468900 c.1625G>A p.G542E Chr.13: 41192633-41192633 on Build GRCh38
167824 c.1636C>T p.R546W Chr.13: 41192622-41192622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754048481 Chr.13: 41192762-41192762 on Build GRCh38
rs760789401 Chr.13: 41192755-41192755 on Build GRCh38
rs975357841 Chr.13: 41192777-41192777 on Build GRCh38
rs745664907 Chr.13: 41192680-41192680 on Build GRCh38
rs753184411 Chr.13: 41192788-41192788 on Build GRCh38
rs149586667 Chr.13: 41192622-41192622 on Build GRCh38
rs199565482 Chr.13: 41192692-41192692 on Build GRCh38
rs373720913 Chr.13: 41192716-41192716 on Build GRCh38
rs757912355 Chr.13: 41192633-41192633 on Build GRCh38
rs770588628 Chr.13: 41192668-41192668 on Build GRCh38
rs143229144 Chr.13: 41192603-41192603 on Build GRCh38
rs975024912 Chr.13: 41192749-41192749 on Build GRCh38
rs987539481 Chr.13: 41192661-41192661 on Build GRCh38
rs773321201 Chr.13: 41192743-41192743 on Build GRCh38
rs746686863 Chr.13: 41192667-41192667 on Build GRCh38
rs765627949 Chr.13: 41192780-41192780 on Build GRCh38
rs71427468 Chr.13: 41192789-41192789 on Build GRCh38
rs762947676 Chr.13: 41192724-41192724 on Build GRCh38
rs201439264 Chr.13: 41192730-41192730 on Build GRCh38
rs754279461 Chr.13: 41192579-41192579 on Build GRCh38
rs189507322 Chr.13: 41192621-41192621 on Build GRCh38
rs912049748 Chr.13: 41192690-41192690 on Build GRCh38
rs766900063 Chr.13: 41192576-41192576 on Build GRCh38
rs200596787 Chr.13: 41192681-41192681 on Build GRCh38
rs780781349 Chr.13: 41192673-41192673 on Build GRCh38
rs777304885 Chr.13: 41192640-41192640 on Build GRCh38
rs766741608 Chr.13: 41192756-41192756 on Build GRCh38
rs770083002 Chr.13: 41192730-41192730 on Build GRCh38
rs760035283 Chr.13: 41192763-41192763 on Build GRCh38
rs756374974 Chr.13: 41192570-41192570 on Build GRCh38

View Full Product Details