Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 13512755-13513016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC38
Entrez Gene ID
126755
Gene Name
leucine rich repeat containing 38
Gene Aliases
-
UniGene
Hs.657356
Gene Chromosome Location
Chr.1: 13474977-13513747 on Build GRCh38
Transcripts
NM_001010847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600687 c.613G>A p.A205T Chr.1: 13512981-13512981 on Build GRCh38
7375594 c.613G>T p.A205S Chr.1: 13512981-13512981 on Build GRCh38
1333431 c.599G>T p.W200L Chr.1: 13512995-13512995 on Build GRCh38
4941569 c.593T>C p.F198S Chr.1: 13513001-13513001 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs956770561 Chr.1: 13512991-13512991 on Build GRCh38
rs1023109726 Chr.1: 13512852-13512852 on Build GRCh38
rs1012722390 Chr.1: 13512937-13512937 on Build GRCh38
rs982484550 Chr.1: 13512972-13512972 on Build GRCh38
rs2990682 Chr.1: 13512887-13512887 on Build GRCh38
rs952073941 Chr.1: 13512794-13512794 on Build GRCh38
rs543437633 Chr.1: 13512817-13512817 on Build GRCh38
rs758281153 Chr.1: 13512936-13512937 on Build GRCh38
rs890730515 Chr.1: 13512829-13512829 on Build GRCh38
rs1024615620 Chr.1: 13512941-13512941 on Build GRCh38
rs760785856 Chr.1: 13512926-13512926 on Build GRCh38
rs984959078 Chr.1: 13512851-13512851 on Build GRCh38
rs766411182 Chr.1: 13512966-13512966 on Build GRCh38
rs944340698 Chr.1: 13512850-13512850 on Build GRCh38
rs1031042631 Chr.1: 13512976-13512976 on Build GRCh38
rs1050687794 Chr.1: 13512824-13512824 on Build GRCh38
rs868283736 Chr.1: 13512925-13512925 on Build GRCh38
rs537476330 Chr.1: 13512960-13512960 on Build GRCh38
rs139328962 Chr.1: 13512924-13512925 on Build GRCh38
rs562444472 Chr.1: 13512869-13512869 on Build GRCh38
rs971348005 Chr.1: 13512953-13512953 on Build GRCh38
rs778790351 Chr.1: 13512922-13512922 on Build GRCh38
rs3056092 Chr.1: 13512939-13512940 on Build GRCh38
rs568449356 Chr.1: 13512905-13512905 on Build GRCh38
rs576983274 Chr.1: 13512872-13512873 on Build GRCh38
rs554612956 Chr.1: 13512853-13512853 on Build GRCh38
rs928862117 Chr.1: 13512904-13512904 on Build GRCh38

View Full Product Details