Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47573022-47573291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KBTBD4
Entrez Gene ID
55709
Gene Name
kelch repeat and BTB domain containing 4
Gene Aliases
BKLHD4, HSPC252
UniGene
Hs.718483
Gene Chromosome Location
Chr.11: 47572197-47579015 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Gene Symbol
PTPMT1
Entrez Gene ID
114971
Gene Name
protein tyrosine phosphatase, mitochondrial 1
Gene Aliases
DUSP23, MOSP, PLIP, PNAS-129
UniGene
Hs.656205
Gene Chromosome Location
Chr.11: 47565430-47573461 on Build GRCh38
Transcripts
NM_001318716.1 NM_001318717.1 NM_001318718.1 NM_001318719.1 NM_001318720.1 NM_001318721.1 NM_001318722.1 NM_001318723.1 NM_001318724.1 NM_001318725.1 NM_016506.6 NM_018095.5 XM_017018009.1 NM_001143984.1 NM_175732.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
927455 c.1244A>G p.Y415C Chr.11: 47573243-47573243 on Build GRCh38
254754 c.1404G>C p.K468N Chr.11: 47573083-47573083 on Build GRCh38
927454 c.1408G>A p.A470T Chr.11: 47573079-47573079 on Build GRCh38
4867892 c.1250A>G p.Q417R Chr.11: 47573285-47573285 on Build GRCh38
4872707 c.1254C>A p.C418* Chr.11: 47573281-47573281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754909672 Chr.11: 47573265-47573265 on Build GRCh38
rs766367041 Chr.11: 47573243-47573243 on Build GRCh38
rs1001981342 Chr.11: 47573244-47573244 on Build GRCh38
rs937649487 Chr.11: 47573063-47573063 on Build GRCh38
rs775934302 Chr.11: 47573094-47573094 on Build GRCh38
rs558994168 Chr.11: 47573222-47573222 on Build GRCh38
rs753594386 Chr.11: 47573105-47573105 on Build GRCh38
rs897496370 Chr.11: 47573131-47573131 on Build GRCh38
rs779958294 Chr.11: 47573048-47573048 on Build GRCh38
rs180799402 Chr.11: 47573185-47573185 on Build GRCh38
rs926394120 Chr.11: 47573061-47573061 on Build GRCh38
rs781565952 Chr.11: 47573109-47573125 on Build GRCh38
rs888010650 Chr.11: 47573068-47573068 on Build GRCh38
rs777822290 Chr.11: 47573067-47573067 on Build GRCh38
rs771044336 Chr.11: 47573080-47573080 on Build GRCh38
rs140618424 Chr.11: 47573079-47573079 on Build GRCh38
rs573433525 Chr.11: 47573130-47573130 on Build GRCh38
rs779018703 Chr.11: 47573194-47573194 on Build GRCh38
rs145669679 Chr.11: 47573173-47573173 on Build GRCh38
rs760881188 Chr.11: 47573099-47573099 on Build GRCh38
rs772442324 Chr.11: 47573093-47573093 on Build GRCh38
rs746726124 Chr.11: 47573049-47573049 on Build GRCh38
rs200689934 Chr.11: 47573081-47573081 on Build GRCh38
rs758372593 Chr.11: 47573149-47573149 on Build GRCh38
rs146486534 Chr.11: 47573215-47573215 on Build GRCh38
rs745888062 Chr.11: 47573195-47573195 on Build GRCh38
rs780938950 Chr.11: 47573175-47573175 on Build GRCh38
rs764926642 Chr.11: 47573235-47573235 on Build GRCh38
rs754683732 Chr.11: 47573174-47573174 on Build GRCh38
rs749395535 Chr.11: 47573077-47573077 on Build GRCh38
rs751505119 Chr.11: 47573254-47573254 on Build GRCh38
rs772635290 Chr.11: 47573207-47573207 on Build GRCh38
rs750736978 Chr.11: 47573240-47573241 on Build GRCh38
rs200102394 Chr.11: 47573163-47573163 on Build GRCh38
rs770062216 Chr.11: 47573057-47573057 on Build GRCh38
rs541644716 Chr.11: 47573092-47573092 on Build GRCh38
rs942278259 Chr.11: 47573074-47573074 on Build GRCh38
rs199567761 Chr.11: 47573241-47573241 on Build GRCh38
rs768943581 Chr.11: 47573214-47573214 on Build GRCh38
rs771134541 Chr.11: 47573187-47573187 on Build GRCh38
rs764908495 Chr.11: 47573140-47573140 on Build GRCh38
rs763838623 Chr.11: 47573104-47573104 on Build GRCh38
rs749941524 Chr.11: 47573146-47573146 on Build GRCh38
rs761122474 Chr.11: 47573210-47573210 on Build GRCh38
rs149097832 Chr.11: 47573236-47573236 on Build GRCh38

View Full Product Details