Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 787351-787602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPUSD1
Entrez Gene ID
113000
Gene Name
RNA pseudouridylate synthase domain containing 1
Gene Aliases
C16orf40, RLUCL
UniGene
Hs.101742
Gene Chromosome Location
Chr.16: 784974-788383 on Build GRCh38
Transcripts
NM_001324086.1 NM_001324410.1 NM_001324411.1 NM_001324412.1 NM_001324413.1 NM_001324414.1 NM_001324415.1 NM_058192.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379994 c.277C>T p.R93W Chr.16: 787383-787383 on Build GRCh38
3691215 c.278G>T p.R93L Chr.16: 787382-787382 on Build GRCh38
4652792 c.169T>C p.C57R Chr.16: 787569-787569 on Build GRCh38
974290 c.225C>T p.C75C Chr.16: 787435-787435 on Build GRCh38
4453496 c.303A>G p.A101A Chr.16: 787357-787357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778642175 Chr.16: 787503-787503 on Build GRCh38
rs765968902 Chr.16: 787388-787388 on Build GRCh38
rs760728656 Chr.16: 787524-787524 on Build GRCh38
rs751974079 Chr.16: 787467-787467 on Build GRCh38
rs776216055 Chr.16: 787530-787530 on Build GRCh38
rs755749313 Chr.16: 787386-787386 on Build GRCh38
rs749942477 Chr.16: 787384-787384 on Build GRCh38
rs1046202206 Chr.16: 787434-787434 on Build GRCh38
rs147488694 Chr.16: 787376-787376 on Build GRCh38
rs771902736 Chr.16: 787514-787514 on Build GRCh38
rs775281067 Chr.16: 787567-787567 on Build GRCh38
rs373291242 Chr.16: 787402-787402 on Build GRCh38
rs916692654 Chr.16: 787546-787546 on Build GRCh38
rs532476380 Chr.16: 787487-787487 on Build GRCh38
rs780239148 Chr.16: 787496-787496 on Build GRCh38
rs774147173 Chr.16: 787438-787438 on Build GRCh38
rs745960697 Chr.16: 787429-787429 on Build GRCh38
rs375981058 Chr.16: 787450-787450 on Build GRCh38
rs770740241 Chr.16: 787546-787546 on Build GRCh38
rs370324017 Chr.16: 787452-787452 on Build GRCh38
rs770329635 Chr.16: 787525-787525 on Build GRCh38
rs900908326 Chr.16: 787515-787515 on Build GRCh38
rs747957798 Chr.16: 787510-787510 on Build GRCh38
rs769643850 Chr.16: 787504-787504 on Build GRCh38
rs113607417 Chr.16: 787550-787550 on Build GRCh38
rs772991283 Chr.16: 787522-787522 on Build GRCh38
rs772261636 Chr.16: 787480-787480 on Build GRCh38
rs766304687 Chr.16: 787544-787544 on Build GRCh38
rs1016478514 Chr.16: 787540-787540 on Build GRCh38
rs750881414 Chr.16: 787538-787538 on Build GRCh38
rs749591998 Chr.16: 787432-787432 on Build GRCh38
rs769970055 Chr.16: 787431-787431 on Build GRCh38
rs990830403 Chr.16: 787545-787545 on Build GRCh38
rs199529893 Chr.16: 787413-787413 on Build GRCh38
rs778345503 Chr.16: 787405-787405 on Build GRCh38
rs763994085 Chr.16: 787377-787377 on Build GRCh38
rs753985410 Chr.16: 787547-787547 on Build GRCh38
rs565775735 Chr.16: 787536-787536 on Build GRCh38
rs768871283 Chr.16: 787499-787499 on Build GRCh38
rs745352237 Chr.16: 787562-787562 on Build GRCh38
rs772728932 Chr.16: 787447-787447 on Build GRCh38
rs374207163 Chr.16: 787441-787441 on Build GRCh38
rs762994824 Chr.16: 787464-787465 on Build GRCh38
rs565262718 Chr.16: 787461-787461 on Build GRCh38
rs752096465 Chr.16: 787410-787410 on Build GRCh38
rs867118496 Chr.16: 787451-787451 on Build GRCh38
rs746041772 Chr.16: 787372-787372 on Build GRCh38
rs777677649 Chr.16: 787551-787551 on Build GRCh38
rs963142263 Chr.16: 787572-787572 on Build GRCh38
rs747249750 Chr.16: 787498-787498 on Build GRCh38
rs1028518519 Chr.16: 787404-787404 on Build GRCh38
rs753423778 Chr.16: 787454-787454 on Build GRCh38
rs761049010 Chr.16: 787579-787579 on Build GRCh38
rs935125578 Chr.16: 787563-787563 on Build GRCh38
rs142183031 Chr.16: 787582-787582 on Build GRCh38
rs762756096 Chr.16: 787568-787568 on Build GRCh38
rs111345102 Chr.16: 787411-787411 on Build GRCh38
rs774267786 Chr.16: 787380-787380 on Build GRCh38
rs949307098 Chr.16: 787474-787474 on Build GRCh38
rs779083867 Chr.16: 787548-787548 on Build GRCh38
rs775961313 Chr.16: 787559-787559 on Build GRCh38
rs768427216 Chr.16: 787435-787435 on Build GRCh38
rs761798168 Chr.16: 787381-787381 on Build GRCh38
rs769471694 Chr.16: 787564-787564 on Build GRCh38
rs547666072 Chr.16: 787534-787534 on Build GRCh38
rs748146320 Chr.16: 787470-787470 on Build GRCh38
rs773631974 Chr.16: 787578-787578 on Build GRCh38
rs983695403 Chr.16: 787387-787387 on Build GRCh38
rs878980150 Chr.16: 787407-787407 on Build GRCh38
rs766933543 Chr.16: 787581-787581 on Build GRCh38
rs976952982 Chr.16: 787394-787394 on Build GRCh38
rs759287402 Chr.16: 787457-787457 on Build GRCh38
rs781759828 Chr.16: 787473-787473 on Build GRCh38
rs769942774 Chr.16: 787375-787375 on Build GRCh38
rs764493187 Chr.16: 787464-787464 on Build GRCh38
rs373949325 Chr.16: 787541-787541 on Build GRCh38
rs148654690 Chr.16: 787390-787390 on Build GRCh38
rs767716811 Chr.16: 787573-787573 on Build GRCh38
rs949256188 Chr.16: 787414-787414 on Build GRCh38
rs775703280 Chr.16: 787554-787559 on Build GRCh38
rs772027333 Chr.16: 787446-787446 on Build GRCh38
rs760727052 Chr.16: 787573-787574 on Build GRCh38
rs941755720 Chr.16: 787439-787439 on Build GRCh38
rs755017143 Chr.16: 787584-787584 on Build GRCh38
rs750982700 Chr.16: 787477-787477 on Build GRCh38
rs766957011 Chr.16: 787382-787382 on Build GRCh38

View Full Product Details