Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 77546249-77546520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROBO2
Entrez Gene ID
6092
Gene Name
roundabout guidance receptor 2
Gene Aliases
SAX3
UniGene
Hs.13305
Gene Chromosome Location
Chr.3: 75906675-77649964 on Build GRCh38
Transcripts
NM_001128929.3 NM_001290039.1 NM_001290040.1 NM_001290065.1 NM_002942.4 XM_011533981.2 XM_011533982.1 XM_011533983.1 XM_011533984.1 XM_011533985.1 XM_017006986.1 XM_017006987.1 XM_017006988.1 XM_017006989.1 XM_017006990.1 XM_017006991.1 XM_017006992.1 XM_017006993.1 XM_017006994.1 XM_017006995.1 XM_017006996.1 XM_017006997.1 XM_017006998.1 XM_017006999.1 XM_017007000.1 XM_017007001.1 XM_017007002.1 XM_017007003.1 XM_017007004.1 XM_017007005.1 XM_017007006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2826906 c.1088G>T p.W363L Chr.3: 77546443-77546443 on Build GRCh38
4400099 c.1037T>A p.F346Y Chr.3: 77546440-77546440 on Build GRCh38
1264461 c.1082delT p.W363fs*48 Chr.3: 77546437-77546437 on Build GRCh38
3733348 c.1012G>A p.D338N Chr.3: 77546367-77546367 on Build GRCh38
4931306 c.938C>T p.P313L Chr.3: 77546341-77546341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760139372 Chr.3: 77546359-77546359 on Build GRCh38
rs528932153 Chr.3: 77546429-77546429 on Build GRCh38
rs779967776 Chr.3: 77546313-77546313 on Build GRCh38
rs766777177 Chr.3: 77546361-77546361 on Build GRCh38
rs774069818 Chr.3: 77546284-77546284 on Build GRCh38
rs774274587 Chr.3: 77546329-77546329 on Build GRCh38
rs777871354 Chr.3: 77546297-77546297 on Build GRCh38
rs199529182 Chr.3: 77546457-77546457 on Build GRCh38
rs371303330 Chr.3: 77546383-77546383 on Build GRCh38
rs771523003 Chr.3: 77546462-77546462 on Build GRCh38
rs760104008 Chr.3: 77546372-77546372 on Build GRCh38
rs749096639 Chr.3: 77546415-77546415 on Build GRCh38
rs186985871 Chr.3: 77546315-77546315 on Build GRCh38
rs765668999 Chr.3: 77546376-77546376 on Build GRCh38
rs142643426 Chr.3: 77546301-77546301 on Build GRCh38
rs375381925 Chr.3: 77546307-77546307 on Build GRCh38
rs757533687 Chr.3: 77546401-77546401 on Build GRCh38
rs1011795942 Chr.3: 77546423-77546423 on Build GRCh38
rs539954814 Chr.3: 77546388-77546388 on Build GRCh38
rs200553597 Chr.3: 77546314-77546314 on Build GRCh38
rs772770981 Chr.3: 77546358-77546358 on Build GRCh38
rs1002917611 Chr.3: 77546280-77546280 on Build GRCh38
rs138063825 Chr.3: 77546290-77546290 on Build GRCh38
rs772755518 Chr.3: 77546481-77546481 on Build GRCh38
rs575736512 Chr.3: 77546389-77546389 on Build GRCh38
rs368780825 Chr.3: 77546355-77546355 on Build GRCh38
rs761239340 Chr.3: 77546341-77546341 on Build GRCh38
rs986191822 Chr.3: 77546385-77546385 on Build GRCh38
rs780055779 Chr.3: 77546402-77546402 on Build GRCh38
rs78123718 Chr.3: 77546344-77546344 on Build GRCh38
rs1041926054 Chr.3: 77546456-77546456 on Build GRCh38
rs146036715 Chr.3: 77546393-77546393 on Build GRCh38
rs778733106 Chr.3: 77546459-77546459 on Build GRCh38
rs975848189 Chr.3: 77546490-77546490 on Build GRCh38
rs754186170 Chr.3: 77546369-77546369 on Build GRCh38
rs200822173 Chr.3: 77546478-77546478 on Build GRCh38
rs758028049 Chr.3: 77546404-77546404 on Build GRCh38

View Full Product Details