Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 238091099-238091372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCLY
Entrez Gene ID
51540
Gene Name
selenocysteine lyase
Gene Aliases
SCL, hSCL
UniGene
Hs.731909
Gene Chromosome Location
Chr.2: 238060924-238099413 on Build GRCh38
Transcripts
NM_016510.5 NR_037904.1

Gene Symbol
UBE2F-SCLY
Entrez Gene ID
100533179
Gene Name
UBE2F-SCLY readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.471785
Gene Chromosome Location
Chr.2: 237966945-238099413 on Build GRCh38
Transcripts
NM_016510.5 NR_037904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188691 c.901A>G p.M301V Chr.2: 238091234-238091234 on Build GRCh38
4092720 c.945G>T p.K315N Chr.2: 238091254-238091254 on Build GRCh38
7084143 c.903G>A p.M301I Chr.2: 238091236-238091236 on Build GRCh38
5225725 c.895A>G p.T299A Chr.2: 238091228-238091228 on Build GRCh38
7084145 c.924G>C p.V308V Chr.2: 238091257-238091257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200211030 Chr.2: 238091189-238091189 on Build GRCh38
rs147924326 Chr.2: 238091301-238091301 on Build GRCh38
rs779007259 Chr.2: 238091230-238091230 on Build GRCh38
rs750869493 Chr.2: 238091183-238091183 on Build GRCh38
rs766712625 Chr.2: 238091306-238091306 on Build GRCh38
rs1048923396 Chr.2: 238091207-238091207 on Build GRCh38
rs144329932 Chr.2: 238091278-238091278 on Build GRCh38
rs142743175 Chr.2: 238091245-238091245 on Build GRCh38
rs776446971 Chr.2: 238091257-238091257 on Build GRCh38
rs763900457 Chr.2: 238091300-238091300 on Build GRCh38
rs764879904 Chr.2: 238091312-238091312 on Build GRCh38
rs776367069 Chr.2: 238091152-238091152 on Build GRCh38
rs752829870 Chr.2: 238091330-238091330 on Build GRCh38
rs376117955 Chr.2: 238091146-238091146 on Build GRCh38
rs2718 Chr.2: 238091292-238091292 on Build GRCh38
rs537547908 Chr.2: 238091280-238091284 on Build GRCh38
rs761129389 Chr.2: 238091302-238091302 on Build GRCh38
rs748142420 Chr.2: 238091232-238091232 on Build GRCh38
rs759469230 Chr.2: 238091268-238091268 on Build GRCh38
rs199772775 Chr.2: 238091234-238091234 on Build GRCh38
rs775945071 Chr.2: 238091121-238091121 on Build GRCh38
rs780777663 Chr.2: 238091205-238091205 on Build GRCh38
rs1025338018 Chr.2: 238091263-238091263 on Build GRCh38
rs965595287 Chr.2: 238091248-238091248 on Build GRCh38
rs779262881 Chr.2: 238091326-238091326 on Build GRCh38
rs113553319 Chr.2: 238091294-238091294 on Build GRCh38
rs368821916 Chr.2: 238091246-238091246 on Build GRCh38
rs756536629 Chr.2: 238091200-238091200 on Build GRCh38
rs531088860 Chr.2: 238091319-238091319 on Build GRCh38
rs1128552 Chr.2: 238091242-238091242 on Build GRCh38
rs775107723 Chr.2: 238091298-238091298 on Build GRCh38
rs868749342 Chr.2: 238091224-238091224 on Build GRCh38
rs949310125 Chr.2: 238091120-238091120 on Build GRCh38
rs543968100 Chr.2: 238091126-238091126 on Build GRCh38
rs994182455 Chr.2: 238091197-238091197 on Build GRCh38
rs372686078 Chr.2: 238091220-238091220 on Build GRCh38
rs149658315 Chr.2: 238091227-238091227 on Build GRCh38

View Full Product Details