Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144431031-144431298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TONSL
Entrez Gene ID
4796
Gene Name
tonsoku-like, DNA repair protein
Gene Aliases
IKBR, NFKBIL2
UniGene
Hs.675285
Gene Chromosome Location
Chr.8: 144428780-144444488 on Build GRCh38
Transcripts
NM_013432.4 XM_011517048.2 XM_011517049.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6859519 c.3774C>G p.N1258K Chr.8: 144431113-144431113 on Build GRCh38
6859520 c.3297C>G p.N1099K Chr.8: 144431113-144431113 on Build GRCh38
4138644 c.3775C>T p.H1259Y Chr.8: 144431112-144431112 on Build GRCh38
6989787 c.3259G>A p.E1087K Chr.8: 144431151-144431151 on Build GRCh38
1097251 c.3326T>C p.L1109P Chr.8: 144431084-144431084 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782055104 Chr.8: 144431066-144431066 on Build GRCh38
rs115362873 Chr.8: 144431254-144431254 on Build GRCh38
rs782460007 Chr.8: 144431085-144431085 on Build GRCh38
rs140904238 Chr.8: 144431185-144431185 on Build GRCh38
rs373594610 Chr.8: 144431069-144431069 on Build GRCh38
rs368544341 Chr.8: 144431192-144431192 on Build GRCh38
rs782192983 Chr.8: 144431149-144431149 on Build GRCh38
rs781937624 Chr.8: 144431058-144431058 on Build GRCh38
rs370640264 Chr.8: 144431073-144431073 on Build GRCh38
rs549258155 Chr.8: 144431270-144431270 on Build GRCh38
rs781833286 Chr.8: 144431095-144431095 on Build GRCh38
rs782161349 Chr.8: 144431237-144431237 on Build GRCh38
rs542273920 Chr.8: 144431168-144431168 on Build GRCh38
rs371792871 Chr.8: 144431125-144431125 on Build GRCh38
rs782733226 Chr.8: 144431090-144431091 on Build GRCh38
rs782563535 Chr.8: 144431094-144431094 on Build GRCh38
rs145066651 Chr.8: 144431089-144431089 on Build GRCh38
rs368364600 Chr.8: 144431118-144431118 on Build GRCh38
rs916929610 Chr.8: 144431194-144431194 on Build GRCh38
rs114298042 Chr.8: 144431249-144431249 on Build GRCh38
rs782674816 Chr.8: 144431130-144431130 on Build GRCh38
rs782249203 Chr.8: 144431124-144431124 on Build GRCh38
rs782286823 Chr.8: 144431167-144431167 on Build GRCh38
rs574845380 Chr.8: 144431082-144431082 on Build GRCh38
rs376930729 Chr.8: 144431072-144431072 on Build GRCh38
rs782655667 Chr.8: 144431123-144431123 on Build GRCh38
rs569775445 Chr.8: 144431155-144431155 on Build GRCh38
rs150083468 Chr.8: 144431223-144431223 on Build GRCh38
rs782312870 Chr.8: 144431157-144431157 on Build GRCh38
rs372748424 Chr.8: 144431193-144431193 on Build GRCh38
rs13258903 Chr.8: 144431275-144431275 on Build GRCh38
rs374717341 Chr.8: 144431090-144431090 on Build GRCh38
rs781877653 Chr.8: 144431083-144431083 on Build GRCh38
rs953856679 Chr.8: 144431103-144431103 on Build GRCh38

View Full Product Details