Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71994370-71994631 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPH2
Entrez Gene ID
121278
Gene Name
tryptophan hydroxylase 2
Gene Aliases
ADHD7, NTPH
UniGene
Hs.736576
Gene Chromosome Location
Chr.12: 71938846-72032441 on Build GRCh38
Transcripts
NM_173353.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
352470 c.1023G>T p.L341L Chr.12: 71994520-71994520 on Build GRCh38
4410450 c.1026G>A p.A342A Chr.12: 71994523-71994523 on Build GRCh38
4398323 c.1007C>T p.S336L Chr.12: 71994504-71994504 on Build GRCh38
109183 c.1034G>A p.G345E Chr.12: 71994531-71994531 on Build GRCh38
3465069 c.973C>T p.P325S Chr.12: 71994470-71994470 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754918661 Chr.12: 71994490-71994490 on Build GRCh38
rs370276646 Chr.12: 71994539-71994539 on Build GRCh38
rs927946801 Chr.12: 71994414-71994414 on Build GRCh38
rs1055143660 Chr.12: 71994467-71994467 on Build GRCh38
rs946786142 Chr.12: 71994497-71994497 on Build GRCh38
rs376192429 Chr.12: 71994432-71994432 on Build GRCh38
rs771288338 Chr.12: 71994420-71994420 on Build GRCh38
rs766153353 Chr.12: 71994481-71994481 on Build GRCh38
rs775596364 Chr.12: 71994443-71994443 on Build GRCh38
rs764304651 Chr.12: 71994458-71994458 on Build GRCh38
rs755764502 Chr.12: 71994522-71994522 on Build GRCh38
rs2887147 Chr.12: 71994480-71994480 on Build GRCh38
rs761981064 Chr.12: 71994590-71994590 on Build GRCh38
rs886049816 Chr.12: 71994547-71994547 on Build GRCh38
rs903580551 Chr.12: 71994484-71994484 on Build GRCh38
rs774911442 Chr.12: 71994431-71994431 on Build GRCh38
rs142857022 Chr.12: 71994531-71994531 on Build GRCh38
rs766171097 Chr.12: 71994592-71994592 on Build GRCh38
rs772478895 Chr.12: 71994541-71994541 on Build GRCh38
rs571084154 Chr.12: 71994591-71994591 on Build GRCh38
rs750159932 Chr.12: 71994469-71994469 on Build GRCh38
rs760757722 Chr.12: 71994564-71994564 on Build GRCh38
rs2887148 Chr.12: 71994478-71994478 on Build GRCh38
rs777289128 Chr.12: 71994496-71994496 on Build GRCh38
rs527300788 Chr.12: 71994435-71994435 on Build GRCh38
rs760839526 Chr.12: 71994455-71994455 on Build GRCh38
rs1007023 Chr.12: 71994594-71994594 on Build GRCh38
rs775508656 Chr.12: 71994548-71994548 on Build GRCh38
rs749678092 Chr.12: 71994413-71994413 on Build GRCh38
rs143153059 Chr.12: 71994514-71994514 on Build GRCh38
rs779187193 Chr.12: 71994538-71994538 on Build GRCh38
rs532719315 Chr.12: 71994565-71994565 on Build GRCh38
rs564050214 Chr.12: 71994523-71994523 on Build GRCh38
rs776828211 Chr.12: 71994581-71994581 on Build GRCh38
rs771325391 Chr.12: 71994532-71994532 on Build GRCh38
rs757480713 Chr.12: 71994557-71994558 on Build GRCh38

View Full Product Details