Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24207301-24207540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
UniGene
Hs.314432
Gene Chromosome Location
Chr.14: 24205524-24208362 on Build GRCh38
Transcripts
NM_001184739.1 NM_001308067.1 NM_174944.3 XM_011536663.2 XM_017021226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2032876 c.678C>T p.G226G Chr.14: 24207383-24207383 on Build GRCh38
5158733 c.763delA p.K256fs*3 Chr.14: 24207438-24207438 on Build GRCh38
5734534 c.758_760delAGG p.E254delE Chr.14: 24207463-24207465 on Build GRCh38
2032877 c.678C>T p.G226G Chr.14: 24207383-24207383 on Build GRCh38
1369328 c.733delA p.K246fs*3 Chr.14: 24207438-24207438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746522000 Chr.14: 24207331-24207331 on Build GRCh38
rs1029490199 Chr.14: 24207369-24207369 on Build GRCh38
rs751669324 Chr.14: 24207368-24207368 on Build GRCh38
rs771147176 Chr.14: 24207467-24207467 on Build GRCh38
rs750254864 Chr.14: 24207389-24207389 on Build GRCh38
rs961300023 Chr.14: 24207399-24207399 on Build GRCh38
rs752772563 Chr.14: 24207375-24207375 on Build GRCh38
rs759823998 Chr.14: 24207476-24207476 on Build GRCh38
rs776266706 Chr.14: 24207337-24207337 on Build GRCh38
rs966429415 Chr.14: 24207364-24207364 on Build GRCh38
rs770166102 Chr.14: 24207348-24207348 on Build GRCh38
rs765064827 Chr.14: 24207384-24207384 on Build GRCh38
rs372943435 Chr.14: 24207457-24207457 on Build GRCh38
rs374020410 Chr.14: 24207514-24207514 on Build GRCh38
rs767643845 Chr.14: 24207374-24207374 on Build GRCh38
rs150354898 Chr.14: 24207370-24207370 on Build GRCh38
rs138176697 Chr.14: 24207490-24207490 on Build GRCh38
rs780886326 Chr.14: 24207423-24207423 on Build GRCh38
rs369574617 Chr.14: 24207383-24207383 on Build GRCh38
rs570224362 Chr.14: 24207336-24207336 on Build GRCh38
rs747637808 Chr.14: 24207434-24207434 on Build GRCh38
rs759003569 Chr.14: 24207394-24207394 on Build GRCh38
rs770352459 Chr.14: 24207444-24207444 on Build GRCh38
rs775598317 Chr.14: 24207504-24207504 on Build GRCh38
rs763173675 Chr.14: 24207363-24207363 on Build GRCh38
rs762476230 Chr.14: 24207347-24207347 on Build GRCh38
rs141252083 Chr.14: 24207352-24207352 on Build GRCh38
rs375900726 Chr.14: 24207513-24207513 on Build GRCh38
rs939226962 Chr.14: 24207447-24207447 on Build GRCh38
rs774688127 Chr.14: 24207473-24207473 on Build GRCh38
rs760698429 Chr.14: 24207506-24207506 on Build GRCh38
rs150827323 Chr.14: 24207365-24207365 on Build GRCh38
rs557459590 Chr.14: 24207509-24207509 on Build GRCh38
rs138195067 Chr.14: 24207407-24207407 on Build GRCh38
rs754671970 Chr.14: 24207412-24207412 on Build GRCh38
rs773548004 Chr.14: 24207453-24207453 on Build GRCh38
rs565378292 Chr.14: 24207395-24207395 on Build GRCh38
rs34031186 Chr.14: 24207521-24207521 on Build GRCh38
rs765252953 Chr.14: 24207508-24207508 on Build GRCh38
rs768089478 Chr.14: 24207477-24207477 on Build GRCh38

View Full Product Details