Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 56496476-56496749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLPP3
Entrez Gene ID
8613
Gene Name
phospholipid phosphatase 3
Gene Aliases
Dri42, LPP3, PAP2B, PPAP2B, VCIP
UniGene
Hs.405156
Gene Chromosome Location
Chr.1: 56494747-56579584 on Build GRCh38
Transcripts
NM_003713.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
276852 c.814T>C p.F272L Chr.1: 56496673-56496673 on Build GRCh38
5381289 c.888T>C p.L296L Chr.1: 56496599-56496599 on Build GRCh38
911056 c.874C>T p.R292W Chr.1: 56496613-56496613 on Build GRCh38
354132 c.841A>T p.K281* Chr.1: 56496646-56496646 on Build GRCh38
6567123 c.831C>G p.L277L Chr.1: 56496656-56496656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746192689 Chr.1: 56496633-56496633 on Build GRCh38
rs150216828 Chr.1: 56496639-56496639 on Build GRCh38
rs761716322 Chr.1: 56496508-56496508 on Build GRCh38
rs540908299 Chr.1: 56496699-56496699 on Build GRCh38
rs763211100 Chr.1: 56496726-56496726 on Build GRCh38
rs201786511 Chr.1: 56496700-56496700 on Build GRCh38
rs755224791 Chr.1: 56496691-56496691 on Build GRCh38
rs1026263580 Chr.1: 56496698-56496698 on Build GRCh38
rs914138911 Chr.1: 56496595-56496595 on Build GRCh38
rs371483426 Chr.1: 56496668-56496668 on Build GRCh38
rs935658742 Chr.1: 56496562-56496562 on Build GRCh38
rs1802022 Chr.1: 56496546-56496546 on Build GRCh38
rs367977475 Chr.1: 56496529-56496529 on Build GRCh38
rs146593813 Chr.1: 56496616-56496616 on Build GRCh38
rs200711664 Chr.1: 56496662-56496662 on Build GRCh38
rs182999910 Chr.1: 56496667-56496667 on Build GRCh38
rs147388964 Chr.1: 56496592-56496592 on Build GRCh38
rs748709096 Chr.1: 56496566-56496566 on Build GRCh38
rs747432529 Chr.1: 56496713-56496713 on Build GRCh38
rs772704217 Chr.1: 56496589-56496589 on Build GRCh38
rs899202466 Chr.1: 56496661-56496661 on Build GRCh38
rs925608207 Chr.1: 56496537-56496537 on Build GRCh38
rs867699788 Chr.1: 56496567-56496567 on Build GRCh38
rs773818825 Chr.1: 56496638-56496638 on Build GRCh38
rs779204041 Chr.1: 56496692-56496692 on Build GRCh38
rs781597987 Chr.1: 56496718-56496718 on Build GRCh38
rs945579050 Chr.1: 56496596-56496596 on Build GRCh38
rs750179615 Chr.1: 56496524-56496524 on Build GRCh38
rs199779037 Chr.1: 56496517-56496517 on Build GRCh38
rs890556930 Chr.1: 56496640-56496640 on Build GRCh38
rs17850456 Chr.1: 56496642-56496642 on Build GRCh38
rs752814070 Chr.1: 56496696-56496696 on Build GRCh38
rs779117993 Chr.1: 56496555-56496555 on Build GRCh38
rs936269531 Chr.1: 56496629-56496629 on Build GRCh38
rs747279885 Chr.1: 56496613-56496613 on Build GRCh38
rs575915994 Chr.1: 56496619-56496619 on Build GRCh38
rs760418890 Chr.1: 56496563-56496565 on Build GRCh38
rs917338023 Chr.1: 56496572-56496572 on Build GRCh38
rs753952934 Chr.1: 56496532-56496532 on Build GRCh38
rs762473321 Chr.1: 56496624-56496624 on Build GRCh38
rs767342556 Chr.1: 56496641-56496641 on Build GRCh38
rs771153064 Chr.1: 56496717-56496717 on Build GRCh38
rs755065427 Chr.1: 56496539-56496539 on Build GRCh38
rs1014113703 Chr.1: 56496702-56496702 on Build GRCh38

View Full Product Details