Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 62358163-62358430 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF11A
Entrez Gene ID
8792
Gene Name
TNF receptor superfamily member 11a
Gene Aliases
CD265, FEO, LOH18CR1, ODFR, OFE, OPTB7, OSTS, PDB2, RANK, TRANCER
UniGene
Hs.204044
Gene Chromosome Location
Chr.18: 62325287-62388096 on Build GRCh38
Transcripts
NM_001270949.1 NM_001270950.1 NM_001270951.1 NM_001278268.1 NM_003839.3 XM_011526244.2 XM_011526245.2 XM_017026064.1 XM_017026065.1 XM_017026066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2881471 c.482A>T p.D161V Chr.18: 62358302-62358302 on Build GRCh38
1324535 c.491C>T p.S164F Chr.18: 62358311-62358311 on Build GRCh38
7246955 c.428-7C>T p.? Chr.18: 62358241-62358241 on Build GRCh38
95375 c.517A>G p.T173A Chr.18: 62358337-62358337 on Build GRCh38
3717974 c.439A>G p.K147E Chr.18: 62358259-62358259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767992510 Chr.18: 62358407-62358407 on Build GRCh38
rs71160827 Chr.18: 62358239-62358240 on Build GRCh38
rs760431356 Chr.18: 62358222-62358223 on Build GRCh38
rs922077544 Chr.18: 62358320-62358320 on Build GRCh38
rs977873572 Chr.18: 62358186-62358186 on Build GRCh38
rs752338757 Chr.18: 62358230-62358230 on Build GRCh38
rs772825793 Chr.18: 62358222-62358224 on Build GRCh38
rs869223834 Chr.18: 62358238-62358240 on Build GRCh38
rs757239013 Chr.18: 62358248-62358248 on Build GRCh38
rs759831655 Chr.18: 62358318-62358318 on Build GRCh38
rs751724442 Chr.18: 62358229-62358230 on Build GRCh38
rs747557225 Chr.18: 62358308-62358308 on Build GRCh38
rs774553906 Chr.18: 62358212-62358212 on Build GRCh38
rs777378404 Chr.18: 62358240-62358240 on Build GRCh38
rs932936700 Chr.18: 62358267-62358268 on Build GRCh38
rs771428401 Chr.18: 62358315-62358315 on Build GRCh38
rs762616067 Chr.18: 62358370-62358370 on Build GRCh38
rs977344921 Chr.18: 62358234-62358234 on Build GRCh38
rs565994027 Chr.18: 62358228-62358228 on Build GRCh38
rs770880146 Chr.18: 62358201-62358201 on Build GRCh38
rs373982231 Chr.18: 62358306-62358306 on Build GRCh38
rs751082710 Chr.18: 62358388-62358388 on Build GRCh38
rs549404558 Chr.18: 62358196-62358196 on Build GRCh38
rs886042486 Chr.18: 62358270-62358270 on Build GRCh38
rs758531470 Chr.18: 62358226-62358226 on Build GRCh38
rs753683719 Chr.18: 62358225-62358226 on Build GRCh38
rs772166671 Chr.18: 62358288-62358288 on Build GRCh38
rs766024657 Chr.18: 62358225-62358225 on Build GRCh38
rs748333316 Chr.18: 62358284-62358284 on Build GRCh38
rs773710150 Chr.18: 62358218-62358218 on Build GRCh38
rs551534851 Chr.18: 62358406-62358406 on Build GRCh38
rs768727205 Chr.18: 62358209-62358209 on Build GRCh38
rs761125787 Chr.18: 62358219-62358219 on Build GRCh38
rs757503631 Chr.18: 62358241-62358241 on Build GRCh38
rs761783046 Chr.18: 62358214-62358214 on Build GRCh38
rs764785953 Chr.18: 62358229-62358229 on Build GRCh38
rs763730028 Chr.18: 62358227-62358227 on Build GRCh38
rs397702236 Chr.18: 62358240-62358241 on Build GRCh38
rs376096275 Chr.18: 62358317-62358317 on Build GRCh38
rs765573888 Chr.18: 62358355-62358355 on Build GRCh38
rs745349765 Chr.18: 62358226-62358227 on Build GRCh38
rs755282653 Chr.18: 62358277-62358277 on Build GRCh38
rs121908655 Chr.18: 62358328-62358328 on Build GRCh38
rs765300869 Chr.18: 62358226-62358228 on Build GRCh38
rs772128691 Chr.18: 62358216-62358216 on Build GRCh38
rs781084597 Chr.18: 62358207-62358207 on Build GRCh38
rs376113816 Chr.18: 62358267-62358267 on Build GRCh38
rs527420709 Chr.18: 62358397-62358397 on Build GRCh38
rs757330552 Chr.18: 62358390-62358390 on Build GRCh38
rs775038704 Chr.18: 62358362-62358362 on Build GRCh38
rs766783108 Chr.18: 62358226-62358226 on Build GRCh38
rs201342790 Chr.18: 62358377-62358377 on Build GRCh38
rs145487506 Chr.18: 62358283-62358283 on Build GRCh38
rs751564829 Chr.18: 62358241-62358241 on Build GRCh38
rs1037002299 Chr.18: 62358400-62358400 on Build GRCh38
rs78551492 Chr.18: 62358227-62358227 on Build GRCh38
rs151031308 Chr.18: 62358319-62358319 on Build GRCh38
rs80232355 Chr.18: 62358239-62358239 on Build GRCh38

View Full Product Details