Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6391288-6391519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD1
Entrez Gene ID
6609
Gene Name
sphingomyelin phosphodiesterase 1
Gene Aliases
ASM, ASMASE, NPD
UniGene
Hs.498173
Gene Chromosome Location
Chr.11: 6390301-6394998 on Build GRCh38
Transcripts
NM_000543.4 NM_001007593.2 NM_001318087.1 NM_001318088.1 NR_027400.2 NR_134502.1 XM_011520303.1 XM_011520304.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3700117 c.367A>T p.N123Y Chr.11: 6391432-6391432 on Build GRCh38
1226894 c.340G>A p.V114M Chr.11: 6391405-6391405 on Build GRCh38
930090 c.417C>A p.L139L Chr.11: 6391482-6391482 on Build GRCh38
4836055 c.452C>G p.S151* Chr.11: 6391517-6391517 on Build GRCh38
3986337 c.433G>A p.V145M Chr.11: 6391498-6391498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757091200 Chr.11: 6391377-6391377 on Build GRCh38
rs1006197646 Chr.11: 6391311-6391311 on Build GRCh38
rs748395897 Chr.11: 6391489-6391489 on Build GRCh38
rs1053828778 Chr.11: 6391331-6391331 on Build GRCh38
rs775743387 Chr.11: 6391393-6391393 on Build GRCh38
rs543145636 Chr.11: 6391401-6391401 on Build GRCh38
rs797044797 Chr.11: 6391481-6391481 on Build GRCh38
rs772349606 Chr.11: 6391392-6391392 on Build GRCh38
rs892451507 Chr.11: 6391334-6391334 on Build GRCh38
rs1057517086 Chr.11: 6391484-6391485 on Build GRCh38
rs747531830 Chr.11: 6391436-6391436 on Build GRCh38
rs780081189 Chr.11: 6391482-6391482 on Build GRCh38
rs769052880 Chr.11: 6391395-6391395 on Build GRCh38
rs371141815 Chr.11: 6391455-6391455 on Build GRCh38
rs149770879 Chr.11: 6391403-6391403 on Build GRCh38
rs767122852 Chr.11: 6391413-6391413 on Build GRCh38
rs779080655 Chr.11: 6391382-6391382 on Build GRCh38
rs906605440 Chr.11: 6391364-6391364 on Build GRCh38
rs879144765 Chr.11: 6391347-6391347 on Build GRCh38
rs142215226 Chr.11: 6391405-6391405 on Build GRCh38
rs923028036 Chr.11: 6391404-6391404 on Build GRCh38
rs556858558 Chr.11: 6391312-6391312 on Build GRCh38
rs140202512 Chr.11: 6391402-6391402 on Build GRCh38
rs867094051 Chr.11: 6391447-6391447 on Build GRCh38
rs59102147 Chr.11: 6391325-6391325 on Build GRCh38
rs370048730 Chr.11: 6391438-6391438 on Build GRCh38
rs148370501 Chr.11: 6391367-6391367 on Build GRCh38
rs763512845 Chr.11: 6391498-6391498 on Build GRCh38
rs768820000 Chr.11: 6391486-6391486 on Build GRCh38
rs202206564 Chr.11: 6391414-6391414 on Build GRCh38
rs747223735 Chr.11: 6391394-6391394 on Build GRCh38
rs773650118 Chr.11: 6391497-6391497 on Build GRCh38
rs756030863 Chr.11: 6391368-6391368 on Build GRCh38
rs560680564 Chr.11: 6391360-6391360 on Build GRCh38
rs777192812 Chr.11: 6391340-6391340 on Build GRCh38
rs755805263 Chr.11: 6391433-6391433 on Build GRCh38
rs189859589 Chr.11: 6391459-6391459 on Build GRCh38
rs745865811 Chr.11: 6391391-6391391 on Build GRCh38
rs752225278 Chr.11: 6391496-6391498 on Build GRCh38
rs747284877 Chr.11: 6391485-6391485 on Build GRCh38
rs375429408 Chr.11: 6391344-6391344 on Build GRCh38
rs778655099 Chr.11: 6391458-6391458 on Build GRCh38
rs749287829 Chr.11: 6391374-6391374 on Build GRCh38
rs958654789 Chr.11: 6391379-6391379 on Build GRCh38
rs1009634178 Chr.11: 6391352-6391352 on Build GRCh38
rs143462566 Chr.11: 6391351-6391351 on Build GRCh38
rs770350207 Chr.11: 6391495-6391495 on Build GRCh38
rs1001589747 Chr.11: 6391362-6391362 on Build GRCh38
rs556385880 Chr.11: 6391471-6391471 on Build GRCh38
rs199913218 Chr.11: 6391408-6391408 on Build GRCh38
rs757974084 Chr.11: 6391500-6391502 on Build GRCh38
rs1006549503 Chr.11: 6391397-6391397 on Build GRCh38
rs727504165 Chr.11: 6391419-6391419 on Build GRCh38
rs754692729 Chr.11: 6391361-6391361 on Build GRCh38
rs1037638471 Chr.11: 6391320-6391320 on Build GRCh38

View Full Product Details