Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2763722-2763949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGTA
Entrez Gene ID
6449
Gene Name
small glutamine rich tetratricopeptide repeat containing alpha
Gene Aliases
SGT, alphaSGT, hSGT
UniGene
Hs.203910
Gene Chromosome Location
Chr.19: 2754714-2783356 on Build GRCh38
Transcripts
NM_003021.3 XM_011528178.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5663710 c.400G>A p.A134T Chr.19: 2763750-2763750 on Build GRCh38
5741339 c.397G>A p.A133T Chr.19: 2763753-2763753 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1029447345 Chr.19: 2763914-2763914 on Build GRCh38
rs760768551 Chr.19: 2763808-2763808 on Build GRCh38
rs994595414 Chr.19: 2763877-2763877 on Build GRCh38
rs572026061 Chr.19: 2763861-2763861 on Build GRCh38
rs1038512360 Chr.19: 2763865-2763865 on Build GRCh38
rs770937649 Chr.19: 2763771-2763771 on Build GRCh38
rs748418318 Chr.19: 2763764-2763768 on Build GRCh38
rs950379275 Chr.19: 2763809-2763809 on Build GRCh38
rs376631698 Chr.19: 2763793-2763793 on Build GRCh38
rs772504422 Chr.19: 2763772-2763773 on Build GRCh38
rs898944967 Chr.19: 2763866-2763866 on Build GRCh38
rs915895071 Chr.19: 2763803-2763803 on Build GRCh38
rs377026231 Chr.19: 2763754-2763754 on Build GRCh38
rs774493814 Chr.19: 2763786-2763786 on Build GRCh38
rs535305854 Chr.19: 2763846-2763846 on Build GRCh38
rs759387077 Chr.19: 2763790-2763790 on Build GRCh38
rs553802828 Chr.19: 2763852-2763852 on Build GRCh38
rs1046025116 Chr.19: 2763851-2763851 on Build GRCh38
rs752931609 Chr.19: 2763806-2763806 on Build GRCh38
rs940171792 Chr.19: 2763855-2763855 on Build GRCh38
rs991937842 Chr.19: 2763799-2763799 on Build GRCh38
rs893362154 Chr.19: 2763834-2763834 on Build GRCh38

View Full Product Details