Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28273959-28274232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SESN2
Entrez Gene ID
83667
Gene Name
sestrin 2
Gene Aliases
HI95, SES2, SEST2
UniGene
Hs.469543
Gene Chromosome Location
Chr.1: 28259452-28282491 on Build GRCh38
Transcripts
NM_031459.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6879040 c.1003C>T p.P335S Chr.1: 28274141-28274141 on Build GRCh38
3677647 c.958A>G p.T320A Chr.1: 28274096-28274096 on Build GRCh38
6858659 c.1007C>A p.T336N Chr.1: 28274145-28274145 on Build GRCh38
2236673 c.982C>A p.R328R Chr.1: 28274120-28274120 on Build GRCh38
23432 c.986G>A p.R329K Chr.1: 28274124-28274124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776026085 Chr.1: 28274130-28274130 on Build GRCh38
rs762021143 Chr.1: 28274143-28274143 on Build GRCh38
rs374949115 Chr.1: 28274044-28274044 on Build GRCh38
rs549821005 Chr.1: 28274102-28274102 on Build GRCh38
rs753443680 Chr.1: 28274104-28274105 on Build GRCh38
rs114922467 Chr.1: 28274063-28274063 on Build GRCh38
rs368191829 Chr.1: 28274151-28274151 on Build GRCh38
rs774059984 Chr.1: 28274002-28274002 on Build GRCh38
rs146457553 Chr.1: 28274133-28274133 on Build GRCh38
rs1016997137 Chr.1: 28274123-28274123 on Build GRCh38
rs762074057 Chr.1: 28274040-28274040 on Build GRCh38
rs763974370 Chr.1: 28274202-28274202 on Build GRCh38
rs1043428602 Chr.1: 28273986-28273986 on Build GRCh38
rs1014337874 Chr.1: 28274073-28274073 on Build GRCh38
rs113713521 Chr.1: 28274140-28274140 on Build GRCh38
rs775873574 Chr.1: 28274013-28274013 on Build GRCh38
rs371949519 Chr.1: 28274005-28274005 on Build GRCh38
rs745325190 Chr.1: 28274115-28274115 on Build GRCh38
rs777523043 Chr.1: 28273992-28273992 on Build GRCh38
rs756176668 Chr.1: 28274201-28274201 on Build GRCh38
rs1048427272 Chr.1: 28274110-28274110 on Build GRCh38
rs374292473 Chr.1: 28274141-28274141 on Build GRCh38
rs764474471 Chr.1: 28274017-28274017 on Build GRCh38
rs368611219 Chr.1: 28273994-28273994 on Build GRCh38
rs1055842387 Chr.1: 28274009-28274009 on Build GRCh38
rs752655380 Chr.1: 28274196-28274196 on Build GRCh38
rs771504819 Chr.1: 28274121-28274121 on Build GRCh38
rs761052546 Chr.1: 28274015-28274015 on Build GRCh38
rs2274848 Chr.1: 28274096-28274096 on Build GRCh38
rs758415728 Chr.1: 28274058-28274058 on Build GRCh38
rs755938790 Chr.1: 28274077-28274077 on Build GRCh38
rs777757853 Chr.1: 28274081-28274081 on Build GRCh38
rs759954285 Chr.1: 28274006-28274006 on Build GRCh38
rs766406851 Chr.1: 28274190-28274190 on Build GRCh38
rs763299927 Chr.1: 28274164-28274164 on Build GRCh38
rs564338338 Chr.1: 28274076-28274076 on Build GRCh38
rs922305536 Chr.1: 28274163-28274163 on Build GRCh38
rs372114042 Chr.1: 28274174-28274174 on Build GRCh38
rs748933770 Chr.1: 28274087-28274087 on Build GRCh38
rs750586812 Chr.1: 28274052-28274052 on Build GRCh38
rs754030508 Chr.1: 28274030-28274030 on Build GRCh38
rs748985099 Chr.1: 28273993-28273993 on Build GRCh38

View Full Product Details