Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5922923-5923196 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGB
Entrez Gene ID
1114
Gene Name
chromogranin B
Gene Aliases
SCG1
UniGene
Hs.516874
Gene Chromosome Location
Chr.20: 5911328-5925361 on Build GRCh38
Transcripts
NM_001819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4561492 c.887G>A p.G296E Chr.20: 5923031-5923031 on Build GRCh38
6017803 c.999G>A p.R333R Chr.20: 5923143-5923143 on Build GRCh38
7338179 c.788C>T p.S263F Chr.20: 5922932-5922932 on Build GRCh38
4099939 c.842C>A p.P281H Chr.20: 5922986-5922986 on Build GRCh38
4563303 c.967G>A p.E323K Chr.20: 5923111-5923111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs936686627 Chr.20: 5923083-5923083 on Build GRCh38
rs777267029 Chr.20: 5923021-5923022 on Build GRCh38
rs755308357 Chr.20: 5922976-5922976 on Build GRCh38
rs765528020 Chr.20: 5923140-5923140 on Build GRCh38
rs978770464 Chr.20: 5923043-5923043 on Build GRCh38
rs897757738 Chr.20: 5923168-5923168 on Build GRCh38
rs931762878 Chr.20: 5923084-5923084 on Build GRCh38
rs369359182 Chr.20: 5922990-5922990 on Build GRCh38
rs959112926 Chr.20: 5922994-5922994 on Build GRCh38
rs776721408 Chr.20: 5923134-5923134 on Build GRCh38
rs370757045 Chr.20: 5923065-5923065 on Build GRCh38
rs750480040 Chr.20: 5922966-5922966 on Build GRCh38
rs868621067 Chr.20: 5923076-5923076 on Build GRCh38
rs775226057 Chr.20: 5922993-5922993 on Build GRCh38
rs779863087 Chr.20: 5923089-5923089 on Build GRCh38
rs774165233 Chr.20: 5923121-5923121 on Build GRCh38
rs748047316 Chr.20: 5923126-5923126 on Build GRCh38
rs780688913 Chr.20: 5923063-5923063 on Build GRCh38
rs376350239 Chr.20: 5923045-5923045 on Build GRCh38
rs267606032 Chr.20: 5923031-5923031 on Build GRCh38
rs769829418 Chr.20: 5922955-5922955 on Build GRCh38
rs113289710 Chr.20: 5923014-5923014 on Build GRCh38
rs777479672 Chr.20: 5922983-5922983 on Build GRCh38
rs139656275 Chr.20: 5923165-5923165 on Build GRCh38
rs745538976 Chr.20: 5923068-5923068 on Build GRCh38
rs759016020 Chr.20: 5923036-5923036 on Build GRCh38
rs1036406459 Chr.20: 5923168-5923170 on Build GRCh38
rs758928695 Chr.20: 5922981-5922981 on Build GRCh38
rs373999382 Chr.20: 5922964-5922964 on Build GRCh38
rs760995504 Chr.20: 5922968-5922968 on Build GRCh38
rs769412851 Chr.20: 5923087-5923087 on Build GRCh38
rs368161388 Chr.20: 5923135-5923135 on Build GRCh38
rs769581505 Chr.20: 5922992-5922992 on Build GRCh38
rs756731449 Chr.20: 5923062-5923062 on Build GRCh38
rs768550071 Chr.20: 5922999-5922999 on Build GRCh38
rs765715328 Chr.20: 5923015-5923015 on Build GRCh38
rs572785094 Chr.20: 5922987-5922987 on Build GRCh38
rs150557526 Chr.20: 5923152-5923152 on Build GRCh38
rs555222601 Chr.20: 5923109-5923109 on Build GRCh38
rs778423183 Chr.20: 5923042-5923042 on Build GRCh38
rs372693764 Chr.20: 5923007-5923007 on Build GRCh38
rs762811546 Chr.20: 5922961-5922961 on Build GRCh38
rs148126308 Chr.20: 5923000-5923000 on Build GRCh38
rs866347352 Chr.20: 5923051-5923051 on Build GRCh38
rs147053014 Chr.20: 5922989-5922989 on Build GRCh38
rs866712045 Chr.20: 5923064-5923064 on Build GRCh38
rs752013541 Chr.20: 5923162-5923162 on Build GRCh38
rs749077565 Chr.20: 5923090-5923090 on Build GRCh38
rs746712482 Chr.20: 5923069-5923077 on Build GRCh38
rs763399470 Chr.20: 5923156-5923156 on Build GRCh38
rs766422129 Chr.20: 5923011-5923011 on Build GRCh38
rs770687576 Chr.20: 5922988-5922988 on Build GRCh38
rs764495609 Chr.20: 5923159-5923159 on Build GRCh38
rs770944731 Chr.20: 5923133-5923133 on Build GRCh38
rs753093809 Chr.20: 5923030-5923030 on Build GRCh38
rs141748366 Chr.20: 5922996-5922996 on Build GRCh38
rs779349247 Chr.20: 5922977-5922977 on Build GRCh38
rs753284059 Chr.20: 5922980-5922980 on Build GRCh38
rs202197795 Chr.20: 5922974-5922974 on Build GRCh38
rs553869356 Chr.20: 5922973-5922973 on Build GRCh38

View Full Product Details