Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 38675600-38675845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE8
Entrez Gene ID
144402
Gene Name
copine 8
Gene Aliases
-
UniGene
Hs.40910
Gene Chromosome Location
Chr.12: 38652200-38908593 on Build GRCh38
Transcripts
NM_153634.2 XM_017018851.1 XM_017018852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772757 c.1412T>A p.V471D Chr.12: 38675737-38675737 on Build GRCh38
6299597 c.1420G>A p.A474T Chr.12: 38675729-38675729 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751707805 Chr.12: 38675772-38675772 on Build GRCh38
rs1024210753 Chr.12: 38675761-38675761 on Build GRCh38
rs371583800 Chr.12: 38675709-38675709 on Build GRCh38
rs191982859 Chr.12: 38675655-38675655 on Build GRCh38
rs1010197371 Chr.12: 38675757-38675757 on Build GRCh38
rs368118379 Chr.12: 38675698-38675698 on Build GRCh38
rs765595982 Chr.12: 38675759-38675759 on Build GRCh38
rs772812928 Chr.12: 38675678-38675678 on Build GRCh38
rs1009589385 Chr.12: 38675803-38675803 on Build GRCh38
rs750428674 Chr.12: 38675749-38675749 on Build GRCh38
rs776141018 Chr.12: 38675700-38675700 on Build GRCh38
rs142785316 Chr.12: 38675768-38675768 on Build GRCh38
rs758934752 Chr.12: 38675760-38675760 on Build GRCh38
rs781546704 Chr.12: 38675809-38675809 on Build GRCh38
rs955890457 Chr.12: 38675718-38675718 on Build GRCh38
rs1033225276 Chr.12: 38675708-38675708 on Build GRCh38
rs746065499 Chr.12: 38675815-38675815 on Build GRCh38
rs367778046 Chr.12: 38675758-38675758 on Build GRCh38
rs757254473 Chr.12: 38675787-38675787 on Build GRCh38
rs760058276 Chr.12: 38675690-38675690 on Build GRCh38

View Full Product Details