Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127804608-127804833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGS
Entrez Gene ID
2356
Gene Name
folylpolyglutamate synthase
Gene Aliases
-
UniGene
Hs.335084
Gene Chromosome Location
Chr.9: 127802858-127814520 on Build GRCh38
Transcripts
NM_001018078.2 NM_001288803.1 NM_004957.5 NR_110170.1 XM_005251864.3 XM_011518439.2 XM_017014565.1 XM_017014566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224503 c.374C>T p.T125M Chr.9: 127804688-127804688 on Build GRCh38
6666624 c.355C>T p.R119* Chr.9: 127804669-127804669 on Build GRCh38
3904304 c.365G>A p.G122D Chr.9: 127804679-127804679 on Build GRCh38
3395585 c.356G>A p.R119Q Chr.9: 127804670-127804670 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754668653 Chr.9: 127804633-127804633 on Build GRCh38
rs1037476616 Chr.9: 127804730-127804730 on Build GRCh38
rs971553520 Chr.9: 127804780-127804780 on Build GRCh38
rs113385716 Chr.9: 127804771-127804771 on Build GRCh38
rs956043088 Chr.9: 127804808-127804808 on Build GRCh38
rs765974935 Chr.9: 127804747-127804747 on Build GRCh38
rs1028217424 Chr.9: 127804750-127804750 on Build GRCh38
rs760359291 Chr.9: 127804712-127804713 on Build GRCh38
rs34570466 Chr.9: 127804743-127804743 on Build GRCh38
rs1047628007 Chr.9: 127804682-127804682 on Build GRCh38
rs200083705 Chr.9: 127804669-127804669 on Build GRCh38
rs577637343 Chr.9: 127804713-127804713 on Build GRCh38
rs76383149 Chr.9: 127804644-127804644 on Build GRCh38
rs770970034 Chr.9: 127804663-127804663 on Build GRCh38
rs745955336 Chr.9: 127804741-127804741 on Build GRCh38
rs753380072 Chr.9: 127804708-127804708 on Build GRCh38
rs369014592 Chr.9: 127804634-127804634 on Build GRCh38
rs746787995 Chr.9: 127804726-127804726 on Build GRCh38
rs139138855 Chr.9: 127804681-127804681 on Build GRCh38
rs777478627 Chr.9: 127804719-127804719 on Build GRCh38
rs1023515365 Chr.9: 127804754-127804754 on Build GRCh38
rs144611973 Chr.9: 127804690-127804690 on Build GRCh38
rs781224107 Chr.9: 127804740-127804740 on Build GRCh38
rs777771570 Chr.9: 127804655-127804655 on Build GRCh38
rs868817861 Chr.9: 127804688-127804688 on Build GRCh38
rs375771978 Chr.9: 127804738-127804738 on Build GRCh38
rs1019786574 Chr.9: 127804805-127804805 on Build GRCh38
rs767983589 Chr.9: 127804707-127804707 on Build GRCh38
rs144149207 Chr.9: 127804743-127804743 on Build GRCh38
rs199965349 Chr.9: 127804711-127804711 on Build GRCh38
rs764428906 Chr.9: 127804689-127804689 on Build GRCh38
rs1005822961 Chr.9: 127804716-127804716 on Build GRCh38
rs758071703 Chr.9: 127804654-127804654 on Build GRCh38
rs770160706 Chr.9: 127804670-127804670 on Build GRCh38
rs1031916130 Chr.9: 127804807-127804807 on Build GRCh38
rs886132949 Chr.9: 127804691-127804691 on Build GRCh38
rs560085760 Chr.9: 127804788-127804788 on Build GRCh38
rs557900013 Chr.9: 127804705-127804705 on Build GRCh38
rs1046238869 Chr.9: 127804711-127804712 on Build GRCh38
rs746977203 Chr.9: 127804656-127804656 on Build GRCh38

View Full Product Details