Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28759804-28760029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM222B
Entrez Gene ID
55731
Gene Name
family with sequence similarity 222 member B
Gene Aliases
C17orf63
UniGene
Hs.564533
Gene Chromosome Location
Chr.17: 28755978-28855036 on Build GRCh38
Transcripts
NM_001077498.2 NM_001288631.1 NM_001288632.1 NM_001288633.1 NM_001288634.1 NM_001288635.1 NM_001288636.1 NM_001288637.1 NM_001288638.1 NM_001288639.1 NM_001288640.1 NM_018182.3 XM_011524986.2 XM_011524987.1 XM_011524988.2 XM_017024831.1 XM_017024832.1 XM_017024833.1 XM_017024834.1 XM_017024835.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198255 c.128C>T p.A43V Chr.17: 28759831-28759831 on Build GRCh38
1198254 c.128C>T p.A43V Chr.17: 28759831-28759831 on Build GRCh38
5740924 c.130G>A p.E44K Chr.17: 28759829-28759829 on Build GRCh38
6464723 c.131A>C p.E44A Chr.17: 28759828-28759828 on Build GRCh38
6464724 c.131A>C p.E44A Chr.17: 28759828-28759828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021458591 Chr.17: 28759945-28759945 on Build GRCh38
rs749759030 Chr.17: 28759859-28759859 on Build GRCh38
rs565571561 Chr.17: 28759944-28759944 on Build GRCh38
rs766545315 Chr.17: 28759940-28759940 on Build GRCh38
rs903985178 Chr.17: 28759870-28759870 on Build GRCh38
rs1032737024 Chr.17: 28759936-28759936 on Build GRCh38
rs998752870 Chr.17: 28759955-28759955 on Build GRCh38
rs979952637 Chr.17: 28759976-28759976 on Build GRCh38
rs866526557 Chr.17: 28759847-28759847 on Build GRCh38
rs1034154283 Chr.17: 28759976-28759976 on Build GRCh38
rs996656229 Chr.17: 28759874-28759874 on Build GRCh38
rs956701911 Chr.17: 28760001-28760001 on Build GRCh38
rs780509836 Chr.17: 28759831-28759831 on Build GRCh38
rs971287306 Chr.17: 28759975-28759975 on Build GRCh38
rs879023578 Chr.17: 28759962-28759962 on Build GRCh38
rs540612554 Chr.17: 28759938-28759938 on Build GRCh38
rs532017770 Chr.17: 28759954-28759954 on Build GRCh38
rs562263244 Chr.17: 28759982-28759982 on Build GRCh38
rs375690040 Chr.17: 28759913-28759913 on Build GRCh38
rs186922550 Chr.17: 28759872-28759872 on Build GRCh38
rs955920432 Chr.17: 28759941-28759941 on Build GRCh38
rs544156777 Chr.17: 28759970-28759970 on Build GRCh38
rs1000364363 Chr.17: 28759881-28759881 on Build GRCh38

View Full Product Details