Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241500342-241500603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FH
Entrez Gene ID
2271
Gene Name
fumarate hydratase
Gene Aliases
FMRD, HLRCC, LRCC, MCL, MCUL1
UniGene
Hs.592490
Gene Chromosome Location
Chr.1: 241497557-241519785 on Build GRCh38
Transcripts
NM_000143.3 XM_011544132.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238897 c.1302C>T p.C434C Chr.1: 241500525-241500525 on Build GRCh38
1296079 c.1357C>G p.L453V Chr.1: 241500470-241500470 on Build GRCh38
6424532 c.1292C>T p.T431I Chr.1: 241500535-241500535 on Build GRCh38
906402 c.1303G>A p.V435M Chr.1: 241500524-241500524 on Build GRCh38
5042966 c.1306G>A p.V436M Chr.1: 241500521-241500521 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766177929 Chr.1: 241500465-241500465 on Build GRCh38
rs201005880 Chr.1: 241500535-241500535 on Build GRCh38
rs201625211 Chr.1: 241500443-241500443 on Build GRCh38
rs763778998 Chr.1: 241500415-241500415 on Build GRCh38
rs201695078 Chr.1: 241500390-241500390 on Build GRCh38
rs398123163 Chr.1: 241500534-241500534 on Build GRCh38
rs863223992 Chr.1: 241500456-241500457 on Build GRCh38
rs200121326 Chr.1: 241500530-241500530 on Build GRCh38
rs863223975 Chr.1: 241500511-241500511 on Build GRCh38
rs774805214 Chr.1: 241500400-241500400 on Build GRCh38
rs749266351 Chr.1: 241500497-241500497 on Build GRCh38
rs772800627 Chr.1: 241500534-241500534 on Build GRCh38
rs992214177 Chr.1: 241500403-241500403 on Build GRCh38
rs201535626 Chr.1: 241500519-241500519 on Build GRCh38
rs201388169 Chr.1: 241500473-241500473 on Build GRCh38
rs140873869 Chr.1: 241500513-241500513 on Build GRCh38
rs863223976 Chr.1: 241500500-241500500 on Build GRCh38
rs147528200 Chr.1: 241500524-241500524 on Build GRCh38
rs876659472 Chr.1: 241500438-241500438 on Build GRCh38
rs201644919 Chr.1: 241500544-241500544 on Build GRCh38
rs758370443 Chr.1: 241500454-241500454 on Build GRCh38
rs398123164 Chr.1: 241500526-241500526 on Build GRCh38
rs863223977 Chr.1: 241500488-241500488 on Build GRCh38
rs1038324354 Chr.1: 241500553-241500553 on Build GRCh38
rs886039367 Chr.1: 241500436-241500436 on Build GRCh38
rs980648661 Chr.1: 241500368-241500368 on Build GRCh38
rs200274147 Chr.1: 241500401-241500401 on Build GRCh38
rs969560424 Chr.1: 241500387-241500387 on Build GRCh38
rs200244096 Chr.1: 241500461-241500461 on Build GRCh38
rs199664825 Chr.1: 241500482-241500482 on Build GRCh38
rs863223989 Chr.1: 241500564-241500564 on Build GRCh38
rs863223990 Chr.1: 241500475-241500478 on Build GRCh38
rs199747704 Chr.1: 241500456-241500456 on Build GRCh38
rs376260044 Chr.1: 241500420-241500420 on Build GRCh38
rs926524019 Chr.1: 241500385-241500385 on Build GRCh38
rs899743834 Chr.1: 241500556-241500556 on Build GRCh38
rs756326120 Chr.1: 241500487-241500487 on Build GRCh38
rs767253363 Chr.1: 241500448-241500448 on Build GRCh38
rs766377516 Chr.1: 241500568-241500568 on Build GRCh38
rs751400708 Chr.1: 241500475-241500475 on Build GRCh38
rs945101483 Chr.1: 241500504-241500504 on Build GRCh38
rs376502356 Chr.1: 241500480-241500480 on Build GRCh38
rs748363858 Chr.1: 241500486-241500486 on Build GRCh38
rs777409218 Chr.1: 241500564-241500564 on Build GRCh38
rs2070080 Chr.1: 241500525-241500525 on Build GRCh38
rs201994824 Chr.1: 241500528-241500528 on Build GRCh38
rs764801019 Chr.1: 241500439-241500439 on Build GRCh38
rs771503401 Chr.1: 241500503-241500503 on Build GRCh38
rs982483795 Chr.1: 241500523-241500523 on Build GRCh38
rs863224009 Chr.1: 241500559-241500559 on Build GRCh38
rs113572177 Chr.1: 241500489-241500489 on Build GRCh38
rs863223991 Chr.1: 241500469-241500470 on Build GRCh38
rs761373027 Chr.1: 241500421-241500421 on Build GRCh38
rs772415507 Chr.1: 241500522-241500522 on Build GRCh38

View Full Product Details