Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27807402-27807645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSG1L
Entrez Gene ID
146395
Gene Name
GSG1 like
Gene Aliases
PRO19651
UniGene
Hs.91910
Gene Chromosome Location
Chr.16: 27787529-28063509 on Build GRCh38
Transcripts
NM_001109763.1 NM_001323900.1 NM_001323901.1 NM_144675.2 XM_011545750.2 XM_011545751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7351805 c.433+1G>T p.? Chr.16: 27807486-27807486 on Build GRCh38
6405719 c.881A>T p.H294L Chr.16: 27807504-27807504 on Build GRCh38
1609143 c.831-9delT p.? Chr.16: 27807563-27807563 on Build GRCh38
1209108 c.898C>T p.R300* Chr.16: 27807487-27807487 on Build GRCh38
6240741 c.462C>T p.D154D Chr.16: 27807512-27807512 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761434979 Chr.16: 27807427-27807427 on Build GRCh38
rs759825867 Chr.16: 27807575-27807575 on Build GRCh38
rs763802879 Chr.16: 27807561-27807561 on Build GRCh38
rs549480929 Chr.16: 27807526-27807526 on Build GRCh38
rs768131186 Chr.16: 27807542-27807542 on Build GRCh38
rs115200908 Chr.16: 27807508-27807508 on Build GRCh38
rs200353968 Chr.16: 27807539-27807539 on Build GRCh38
rs367643686 Chr.16: 27807566-27807566 on Build GRCh38
rs760887097 Chr.16: 27807548-27807548 on Build GRCh38
rs940676851 Chr.16: 27807440-27807440 on Build GRCh38
rs774935373 Chr.16: 27807464-27807464 on Build GRCh38
rs766923159 Chr.16: 27807463-27807463 on Build GRCh38
rs770113472 Chr.16: 27807518-27807518 on Build GRCh38
rs755895622 Chr.16: 27807486-27807486 on Build GRCh38
rs776051779 Chr.16: 27807546-27807546 on Build GRCh38
rs770879565 Chr.16: 27807533-27807533 on Build GRCh38
rs769878553 Chr.16: 27807456-27807456 on Build GRCh38
rs996197799 Chr.16: 27807434-27807434 on Build GRCh38
rs201172768 Chr.16: 27807538-27807538 on Build GRCh38
rs533846776 Chr.16: 27807580-27807580 on Build GRCh38
rs756813833 Chr.16: 27807567-27807567 on Build GRCh38
rs562822713 Chr.16: 27807532-27807532 on Build GRCh38
rs764431711 Chr.16: 27807562-27807563 on Build GRCh38
rs745756770 Chr.16: 27807505-27807505 on Build GRCh38
rs760014713 Chr.16: 27807471-27807471 on Build GRCh38
rs373838978 Chr.16: 27807516-27807516 on Build GRCh38
rs376701649 Chr.16: 27807597-27807597 on Build GRCh38
rs757960551 Chr.16: 27807571-27807572 on Build GRCh38
rs991050818 Chr.16: 27807599-27807599 on Build GRCh38
rs977157553 Chr.16: 27807569-27807570 on Build GRCh38
rs779863070 Chr.16: 27807512-27807512 on Build GRCh38
rs527602928 Chr.16: 27807503-27807503 on Build GRCh38
rs753399352 Chr.16: 27807492-27807492 on Build GRCh38
rs778391502 Chr.16: 27807563-27807566 on Build GRCh38
rs763637151 Chr.16: 27807487-27807487 on Build GRCh38
rs540441054 Chr.16: 27807566-27807567 on Build GRCh38
rs372065461 Chr.16: 27807450-27807450 on Build GRCh38
rs373880812 Chr.16: 27807584-27807584 on Build GRCh38
rs778941623 Chr.16: 27807502-27807502 on Build GRCh38
rs754096851 Chr.16: 27807563-27807563 on Build GRCh38
rs771812987 Chr.16: 27807444-27807444 on Build GRCh38
rs202218910 Chr.16: 27807457-27807457 on Build GRCh38
rs1011036712 Chr.16: 27807525-27807525 on Build GRCh38
rs761242125 Chr.16: 27807521-27807522 on Build GRCh38
rs756651543 Chr.16: 27807497-27807497 on Build GRCh38
rs369998202 Chr.16: 27807478-27807478 on Build GRCh38

View Full Product Details