Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205523285-205523538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3482222 c.129G>A p.E43E Chr.1: 205523296-205523296 on Build GRCh38
1661085 c.164C>T p.P55L Chr.1: 205523516-205523516 on Build GRCh38
3789389 c.168G>A p.Q56Q Chr.1: 205523520-205523520 on Build GRCh38
3482224 c.161C>T p.P54L Chr.1: 205523513-205523513 on Build GRCh38
99053 c.166C>T p.Q56* Chr.1: 205523518-205523518 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200813019 Chr.1: 205523502-205523502 on Build GRCh38
rs750206182 Chr.1: 205523498-205523498 on Build GRCh38
rs753136425 Chr.1: 205523511-205523511 on Build GRCh38
rs776036243 Chr.1: 205523491-205523491 on Build GRCh38
rs1029636704 Chr.1: 205523457-205523457 on Build GRCh38
rs772199728 Chr.1: 205523373-205523373 on Build GRCh38
rs370754957 Chr.1: 205523314-205523314 on Build GRCh38
rs753805512 Chr.1: 205523519-205523519 on Build GRCh38
rs757093372 Chr.1: 205523412-205523412 on Build GRCh38
rs752160165 Chr.1: 205523387-205523387 on Build GRCh38
rs752207911 Chr.1: 205523353-205523353 on Build GRCh38
rs776894776 Chr.1: 205523454-205523454 on Build GRCh38
rs748222295 Chr.1: 205523421-205523421 on Build GRCh38
rs546073701 Chr.1: 205523406-205523406 on Build GRCh38
rs1033972477 Chr.1: 205523461-205523461 on Build GRCh38
rs1039553315 Chr.1: 205523414-205523414 on Build GRCh38
rs759899746 Chr.1: 205523456-205523456 on Build GRCh38
rs532476211 Chr.1: 205523386-205523386 on Build GRCh38
rs754560252 Chr.1: 205523343-205523343 on Build GRCh38
rs754395959 Chr.1: 205523337-205523337 on Build GRCh38
rs1010443645 Chr.1: 205523468-205523468 on Build GRCh38
rs751246855 Chr.1: 205523372-205523372 on Build GRCh38
rs1033072380 Chr.1: 205523352-205523369 on Build GRCh38
rs371344279 Chr.1: 205523475-205523475 on Build GRCh38
rs757040504 Chr.1: 205523360-205523360 on Build GRCh38
rs186264542 Chr.1: 205523313-205523313 on Build GRCh38
rs751181313 Chr.1: 205523407-205523407 on Build GRCh38
rs541452253 Chr.1: 205523361-205523361 on Build GRCh38
rs761066008 Chr.1: 205523324-205523324 on Build GRCh38
rs766298796 Chr.1: 205523318-205523318 on Build GRCh38
rs574489048 Chr.1: 205523366-205523366 on Build GRCh38
rs1041973378 Chr.1: 205523476-205523476 on Build GRCh38
rs767883519 Chr.1: 205523415-205523416 on Build GRCh38
rs182526430 Chr.1: 205523511-205523511 on Build GRCh38
rs768587858 Chr.1: 205523311-205523311 on Build GRCh38
rs746862453 Chr.1: 205523359-205523359 on Build GRCh38
rs755913151 Chr.1: 205523516-205523516 on Build GRCh38
rs769722970 Chr.1: 205523367-205523367 on Build GRCh38
rs779986647 Chr.1: 205523420-205523420 on Build GRCh38
rs757988027 Chr.1: 205523355-205523355 on Build GRCh38
rs756011675 Chr.1: 205523417-205523417 on Build GRCh38
rs748194431 Chr.1: 205523372-205523372 on Build GRCh38
rs543414279 Chr.1: 205523371-205523371 on Build GRCh38
rs774326622 Chr.1: 205523369-205523369 on Build GRCh38
rs569695440 Chr.1: 205523517-205523517 on Build GRCh38
rs191558863 Chr.1: 205523467-205523467 on Build GRCh38
rs773557656 Chr.1: 205523374-205523374 on Build GRCh38
rs945890994 Chr.1: 205523333-205523333 on Build GRCh38
rs757910654 Chr.1: 205523390-205523390 on Build GRCh38
rs35134237 Chr.1: 205523494-205523494 on Build GRCh38
rs778305473 Chr.1: 205523347-205523347 on Build GRCh38
rs766824330 Chr.1: 205523375-205523375 on Build GRCh38
rs772243829 Chr.1: 205523436-205523436 on Build GRCh38
rs750824496 Chr.1: 205523320-205523320 on Build GRCh38
rs996846376 Chr.1: 205523444-205523444 on Build GRCh38
rs755289020 Chr.1: 205523416-205523416 on Build GRCh38
rs559325623 Chr.1: 205523416-205523416 on Build GRCh38
rs777083241 Chr.1: 205523381-205523381 on Build GRCh38
rs773631275 Chr.1: 205523438-205523438 on Build GRCh38
rs767053679 Chr.1: 205523332-205523332 on Build GRCh38
rs563591627 Chr.1: 205523385-205523385 on Build GRCh38
rs74141238 Chr.1: 205523358-205523358 on Build GRCh38
rs763086197 Chr.1: 205523305-205523305 on Build GRCh38
rs200845295 Chr.1: 205523493-205523493 on Build GRCh38
rs747216878 Chr.1: 205523447-205523447 on Build GRCh38

View Full Product Details