Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38841342-38841609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf98
Entrez Gene ID
388381
Gene Name
chromosome 17 open reading frame 98
Gene Aliases
-
UniGene
Hs.250182
Gene Chromosome Location
Chr.17: 38835088-38841389 on Build GRCh38
Transcripts
NM_001080465.2

SNP Detail

SNP ID Chromosome Location
rs901255147 Chr.17: 38841570-38841570 on Build GRCh38
rs570022376 Chr.17: 38841422-38841422 on Build GRCh38
rs553857905 Chr.17: 38841482-38841482 on Build GRCh38
rs772698212 Chr.17: 38841416-38841416 on Build GRCh38
rs543747766 Chr.17: 38841533-38841533 on Build GRCh38
rs773747768 Chr.17: 38841417-38841417 on Build GRCh38
rs998685134 Chr.17: 38841546-38841546 on Build GRCh38
rs1008492486 Chr.17: 38841562-38841562 on Build GRCh38
rs748719541 Chr.17: 38841405-38841405 on Build GRCh38
rs748862711 Chr.17: 38841385-38841385 on Build GRCh38
rs1042955954 Chr.17: 38841493-38841493 on Build GRCh38
rs750179754 Chr.17: 38841380-38841380 on Build GRCh38
rs996894933 Chr.17: 38841578-38841578 on Build GRCh38
rs773857323 Chr.17: 38841407-38841408 on Build GRCh38
rs946588168 Chr.17: 38841497-38841497 on Build GRCh38
rs771211013 Chr.17: 38841423-38841423 on Build GRCh38
rs748901949 Chr.17: 38841485-38841485 on Build GRCh38
rs183629463 Chr.17: 38841550-38841550 on Build GRCh38
rs764046819 Chr.17: 38841365-38841365 on Build GRCh38
rs779524780 Chr.17: 38841384-38841384 on Build GRCh38
rs908798211 Chr.17: 38841415-38841415 on Build GRCh38
rs779578988 Chr.17: 38841396-38841396 on Build GRCh38
rs192519034 Chr.17: 38841468-38841468 on Build GRCh38
rs370498413 Chr.17: 38841395-38841395 on Build GRCh38
rs747606930 Chr.17: 38841382-38841382 on Build GRCh38
rs938475604 Chr.17: 38841467-38841467 on Build GRCh38
rs597234 Chr.17: 38841426-38841426 on Build GRCh38
rs201466719 Chr.17: 38841372-38841372 on Build GRCh38
rs190263024 Chr.17: 38841457-38841457 on Build GRCh38
rs1018576863 Chr.17: 38841574-38841574 on Build GRCh38
rs952335622 Chr.17: 38841400-38841400 on Build GRCh38
rs757056522 Chr.17: 38841374-38841374 on Build GRCh38
rs554864445 Chr.17: 38841510-38841510 on Build GRCh38
rs141665212 Chr.17: 38841443-38841443 on Build GRCh38
rs772454354 Chr.17: 38841488-38841488 on Build GRCh38
rs894112380 Chr.17: 38841474-38841474 on Build GRCh38
rs984336890 Chr.17: 38841401-38841401 on Build GRCh38
rs781022421 Chr.17: 38841379-38841379 on Build GRCh38
rs584121 Chr.17: 38841583-38841583 on Build GRCh38
rs576409280 Chr.17: 38841516-38841516 on Build GRCh38
rs542857859 Chr.17: 38841500-38841500 on Build GRCh38
rs572240185 Chr.17: 38841496-38841496 on Build GRCh38

View Full Product Details