Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23379310-23379561 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM5
Entrez Gene ID
116173
Gene Name
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases
CKLFSF5
UniGene
Hs.99272
Gene Chromosome Location
Chr.14: 23376439-23379772 on Build GRCh38
Transcripts
NM_001037288.1 NM_001288744.1 NM_001288745.1 NM_001288746.1 NM_138460.2 XM_017020953.1 XM_017020954.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
954702 c.646G>A p.G216R Chr.14: 23379371-23379371 on Build GRCh38
6630981 c.449C>T p.A150V Chr.14: 23379375-23379375 on Build GRCh38
2031504 c.399C>A p.I133I Chr.14: 23379325-23379325 on Build GRCh38
4402412 c.458G>A p.G153E Chr.14: 23379474-23379474 on Build GRCh38
954701 c.445G>A p.G149R Chr.14: 23379371-23379371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772534794 Chr.14: 23379476-23379476 on Build GRCh38
rs773998041 Chr.14: 23379468-23379468 on Build GRCh38
rs763065340 Chr.14: 23379504-23379504 on Build GRCh38
rs373158495 Chr.14: 23379383-23379383 on Build GRCh38
rs769668846 Chr.14: 23379336-23379336 on Build GRCh38
rs767389342 Chr.14: 23379508-23379508 on Build GRCh38
rs373722799 Chr.14: 23379516-23379516 on Build GRCh38
rs900092904 Chr.14: 23379450-23379450 on Build GRCh38
rs140631314 Chr.14: 23379353-23379353 on Build GRCh38
rs377511630 Chr.14: 23379440-23379440 on Build GRCh38
rs879488667 Chr.14: 23379528-23379528 on Build GRCh38
rs146554548 Chr.14: 23379370-23379370 on Build GRCh38
rs533328493 Chr.14: 23379447-23379447 on Build GRCh38
rs746834889 Chr.14: 23379528-23379528 on Build GRCh38
rs748983924 Chr.14: 23379374-23379374 on Build GRCh38
rs139534097 Chr.14: 23379500-23379500 on Build GRCh38
rs780804730 Chr.14: 23379430-23379430 on Build GRCh38
rs775685509 Chr.14: 23379478-23379478 on Build GRCh38
rs776085461 Chr.14: 23379390-23379390 on Build GRCh38
rs754829904 Chr.14: 23379423-23379423 on Build GRCh38
rs770567891 Chr.14: 23379375-23379375 on Build GRCh38
rs757607180 Chr.14: 23379402-23379402 on Build GRCh38
rs750603946 Chr.14: 23379415-23379415 on Build GRCh38
rs367985105 Chr.14: 23379482-23379482 on Build GRCh38
rs754669344 Chr.14: 23379334-23379334 on Build GRCh38
rs141394322 Chr.14: 23379479-23379479 on Build GRCh38
rs777216279 Chr.14: 23379487-23379487 on Build GRCh38
rs753710608 Chr.14: 23379539-23379539 on Build GRCh38
rs770369986 Chr.14: 23379465-23379465 on Build GRCh38
rs750682773 Chr.14: 23379502-23379502 on Build GRCh38
rs761061722 Chr.14: 23379396-23379396 on Build GRCh38
rs970565487 Chr.14: 23379517-23379517 on Build GRCh38
rs150499732 Chr.14: 23379354-23379354 on Build GRCh38
rs376066775 Chr.14: 23379538-23379538 on Build GRCh38
rs745460272 Chr.14: 23379471-23379471 on Build GRCh38
rs748783789 Chr.14: 23379455-23379455 on Build GRCh38
rs765364256 Chr.14: 23379501-23379501 on Build GRCh38
rs779214295 Chr.14: 23379513-23379516 on Build GRCh38
rs35045021 Chr.14: 23379374-23379375 on Build GRCh38
rs1009405908 Chr.14: 23379362-23379362 on Build GRCh38
rs1000814044 Chr.14: 23379451-23379451 on Build GRCh38
rs371491921 Chr.14: 23379401-23379401 on Build GRCh38
rs767708625 Chr.14: 23379388-23379388 on Build GRCh38
rs772690985 Chr.14: 23379371-23379371 on Build GRCh38
rs192189914 Chr.14: 23379397-23379397 on Build GRCh38
rs374734156 Chr.14: 23379404-23379404 on Build GRCh38
rs984503112 Chr.14: 23379472-23379472 on Build GRCh38
rs529320104 Chr.14: 23379439-23379439 on Build GRCh38
rs752434162 Chr.14: 23379509-23379509 on Build GRCh38

View Full Product Details