Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78219153-78219426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB2
Entrez Gene ID
9846
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
UniGene
Hs.429434
Gene Chromosome Location
Chr.11: 78215290-78417822 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

Gene Symbol
USP35
Entrez Gene ID
57558
Gene Name
ubiquitin specific peptidase 35
Gene Aliases
-
UniGene
Hs.531249
Gene Chromosome Location
Chr.11: 78188812-78233651 on Build GRCh38
Transcripts
NM_012296.3 NM_080491.2 XM_006718753.2 XM_011545408.2 XM_017018052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
429916 c.2016G>C p.K672N Chr.11: 78219287-78219287 on Build GRCh38
5640611 c.2003C>T p.S668L Chr.11: 78219300-78219300 on Build GRCh38
3752760 c.2004A>G p.S668S Chr.11: 78219299-78219299 on Build GRCh38
6838057 c.1892C>G p.S631C Chr.11: 78219411-78219411 on Build GRCh38
4036924 c.2016G>T p.K672N Chr.11: 78219287-78219287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753285605 Chr.11: 78219294-78219294 on Build GRCh38
rs750329481 Chr.11: 78219277-78219277 on Build GRCh38
rs878921442 Chr.11: 78219197-78219197 on Build GRCh38
rs764153129 Chr.11: 78219233-78219233 on Build GRCh38
rs760617029 Chr.11: 78219362-78219362 on Build GRCh38
rs767246566 Chr.11: 78219358-78219358 on Build GRCh38
rs199861462 Chr.11: 78219232-78219232 on Build GRCh38
rs761708659 Chr.11: 78219357-78219357 on Build GRCh38
rs758024914 Chr.11: 78219391-78219391 on Build GRCh38
rs770369313 Chr.11: 78219318-78219318 on Build GRCh38
rs768976090 Chr.11: 78219348-78219348 on Build GRCh38
rs369763933 Chr.11: 78219274-78219274 on Build GRCh38
rs898721891 Chr.11: 78219282-78219282 on Build GRCh38
rs925182130 Chr.11: 78219199-78219199 on Build GRCh38
rs778418186 Chr.11: 78219309-78219309 on Build GRCh38
rs948039263 Chr.11: 78219196-78219196 on Build GRCh38
rs961085514 Chr.11: 78219328-78219328 on Build GRCh38
rs61749244 Chr.11: 78219401-78219401 on Build GRCh38
rs1052190296 Chr.11: 78219218-78219218 on Build GRCh38
rs759733560 Chr.11: 78219228-78219228 on Build GRCh38
rs752327819 Chr.11: 78219390-78219390 on Build GRCh38
rs180972993 Chr.11: 78219174-78219174 on Build GRCh38
rs923042940 Chr.11: 78219234-78219234 on Build GRCh38
rs115203824 Chr.11: 78219356-78219356 on Build GRCh38
rs144443221 Chr.11: 78219398-78219398 on Build GRCh38
rs779560386 Chr.11: 78219289-78219289 on Build GRCh38
rs114076515 Chr.11: 78219259-78219259 on Build GRCh38
rs753406598 Chr.11: 78219373-78219373 on Build GRCh38
rs530915833 Chr.11: 78219229-78219229 on Build GRCh38
rs1005510814 Chr.11: 78219217-78219217 on Build GRCh38
rs746125014 Chr.11: 78219204-78219204 on Build GRCh38
rs565178188 Chr.11: 78219203-78219203 on Build GRCh38
rs747450985 Chr.11: 78219310-78219310 on Build GRCh38
rs374761381 Chr.11: 78219197-78219203 on Build GRCh38
rs764981508 Chr.11: 78219380-78219380 on Build GRCh38
rs1006282561 Chr.11: 78219278-78219278 on Build GRCh38
rs781659629 Chr.11: 78219313-78219313 on Build GRCh38
rs771169575 Chr.11: 78219312-78219312 on Build GRCh38
rs933045615 Chr.11: 78219258-78219258 on Build GRCh38
rs2450122 Chr.11: 78219299-78219299 on Build GRCh38
rs368153717 Chr.11: 78219210-78219216 on Build GRCh38
rs138587090 Chr.11: 78219281-78219281 on Build GRCh38
rs115982223 Chr.11: 78219332-78219332 on Build GRCh38
rs766302085 Chr.11: 78219370-78219370 on Build GRCh38
rs145964223 Chr.11: 78219314-78219314 on Build GRCh38
rs374489905 Chr.11: 78219247-78219247 on Build GRCh38
rs114715848 Chr.11: 78219374-78219374 on Build GRCh38
rs771527647 Chr.11: 78219223-78219223 on Build GRCh38
rs776982504 Chr.11: 78219226-78219226 on Build GRCh38
rs746171817 Chr.11: 78219400-78219400 on Build GRCh38
rs761496755 Chr.11: 78219266-78219266 on Build GRCh38
rs772941154 Chr.11: 78219360-78219360 on Build GRCh38
rs201966341 Chr.11: 78219325-78219325 on Build GRCh38
rs775714252 Chr.11: 78219319-78219319 on Build GRCh38

View Full Product Details