Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24889299-24889544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPJ
Entrez Gene ID
55835
Gene Name
centromere protein J
Gene Aliases
BM032, CENP-J, CPAP, LAP, LIP1, MCPH6, SASS4, SCKL4, Sas-4
UniGene
Hs.513379
Gene Chromosome Location
Chr.13: 24882274-24922889 on Build GRCh38
Transcripts
NM_018451.4 NR_047594.1 NR_047595.1 XM_011535149.2 XM_011535150.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1238487 c.3221C>T p.T1074I Chr.13: 24889396-24889396 on Build GRCh38
6042733 c.3218A>C p.N1073T Chr.13: 24889399-24889399 on Build GRCh38
3981973 c.3228T>G p.V1076V Chr.13: 24889389-24889389 on Build GRCh38
1366048 c.3223T>C p.S1075P Chr.13: 24889394-24889394 on Build GRCh38
3376519 c.3283A>T p.N1095Y Chr.13: 24889334-24889334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777475321 Chr.13: 24889322-24889322 on Build GRCh38
rs994160275 Chr.13: 24889410-24889410 on Build GRCh38
rs747494411 Chr.13: 24889341-24889342 on Build GRCh38
rs764426666 Chr.13: 24889395-24889395 on Build GRCh38
rs147768793 Chr.13: 24889380-24889380 on Build GRCh38
rs770506371 Chr.13: 24889333-24889333 on Build GRCh38
rs1007704062 Chr.13: 24889516-24889516 on Build GRCh38
rs762029402 Chr.13: 24889431-24889431 on Build GRCh38
rs768726225 Chr.13: 24889349-24889349 on Build GRCh38
rs774578193 Chr.13: 24889407-24889407 on Build GRCh38
rs770063241 Chr.13: 24889419-24889421 on Build GRCh38
rs890692518 Chr.13: 24889453-24889453 on Build GRCh38
rs914789361 Chr.13: 24889382-24889382 on Build GRCh38
rs776285097 Chr.13: 24889343-24889343 on Build GRCh38
rs772108511 Chr.13: 24889482-24889482 on Build GRCh38
rs767547819 Chr.13: 24889436-24889436 on Build GRCh38
rs949026330 Chr.13: 24889468-24889468 on Build GRCh38
rs775902128 Chr.13: 24889387-24889387 on Build GRCh38
rs776699474 Chr.13: 24889360-24889360 on Build GRCh38
rs61739722 Chr.13: 24889328-24889328 on Build GRCh38
rs763373509 Chr.13: 24889388-24889388 on Build GRCh38
rs556953003 Chr.13: 24889330-24889330 on Build GRCh38
rs199422203 Chr.13: 24889371-24889374 on Build GRCh38
rs746336237 Chr.13: 24889350-24889350 on Build GRCh38
rs1052007548 Chr.13: 24889432-24889432 on Build GRCh38
rs897793933 Chr.13: 24889396-24889396 on Build GRCh38

View Full Product Details