Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180908717-180908976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696843 c.209G>A p.G70E Chr.5: 180908745-180908745 on Build GRCh38
1696844 c.209G>A p.G70E Chr.5: 180908745-180908745 on Build GRCh38
4585793 c.356A>C p.Q119P Chr.5: 180908892-180908892 on Build GRCh38
1066874 c.319T>A p.L107M Chr.5: 180908855-180908855 on Build GRCh38
3012759 c.331C>T p.L111F Chr.5: 180908867-180908867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139672035 Chr.5: 180908744-180908744 on Build GRCh38
rs888468555 Chr.5: 180908821-180908821 on Build GRCh38
rs760651856 Chr.5: 180908911-180908911 on Build GRCh38
rs1023220185 Chr.5: 180908907-180908907 on Build GRCh38
rs774068727 Chr.5: 180908850-180908850 on Build GRCh38
rs144634509 Chr.5: 180908813-180908813 on Build GRCh38
rs753356741 Chr.5: 180908889-180908889 on Build GRCh38
rs561171905 Chr.5: 180908871-180908871 on Build GRCh38
rs767424893 Chr.5: 180908806-180908806 on Build GRCh38
rs751937565 Chr.5: 180908902-180908902 on Build GRCh38
rs982188212 Chr.5: 180908878-180908878 on Build GRCh38
rs17141274 Chr.5: 180908815-180908815 on Build GRCh38
rs752267907 Chr.5: 180908810-180908810 on Build GRCh38
rs1041955738 Chr.5: 180908768-180908768 on Build GRCh38
rs763625609 Chr.5: 180908887-180908887 on Build GRCh38
rs970739082 Chr.5: 180908948-180908948 on Build GRCh38
rs112784813 Chr.5: 180908743-180908743 on Build GRCh38
rs367763125 Chr.5: 180908771-180908771 on Build GRCh38
rs973632762 Chr.5: 180908945-180908945 on Build GRCh38
rs563026665 Chr.5: 180908846-180908846 on Build GRCh38
rs865959616 Chr.5: 180908755-180908755 on Build GRCh38
rs529094290 Chr.5: 180908874-180908874 on Build GRCh38
rs573486977 Chr.5: 180908784-180908784 on Build GRCh38
rs747377512 Chr.5: 180908819-180908819 on Build GRCh38
rs748602705 Chr.5: 180908823-180908823 on Build GRCh38
rs962145349 Chr.5: 180908929-180908929 on Build GRCh38
rs764929768 Chr.5: 180908900-180908900 on Build GRCh38
rs371673914 Chr.5: 180908865-180908865 on Build GRCh38
rs760188256 Chr.5: 180908789-180908789 on Build GRCh38
rs747500534 Chr.5: 180908751-180908751 on Build GRCh38
rs770209825 Chr.5: 180908765-180908765 on Build GRCh38
rs763120109 Chr.5: 180908843-180908843 on Build GRCh38
rs751332966 Chr.5: 180908789-180908789 on Build GRCh38
rs759437011 Chr.5: 180908797-180908797 on Build GRCh38
rs564122210 Chr.5: 180908814-180908814 on Build GRCh38
rs761973449 Chr.5: 180908762-180908762 on Build GRCh38
rs1037121173 Chr.5: 180908827-180908827 on Build GRCh38
rs767230646 Chr.5: 180908872-180908872 on Build GRCh38
rs770010555 Chr.5: 180908825-180908825 on Build GRCh38
rs760555544 Chr.5: 180908875-180908875 on Build GRCh38
rs145704337 Chr.5: 180908822-180908822 on Build GRCh38
rs1015063291 Chr.5: 180908912-180908912 on Build GRCh38
rs769346844 Chr.5: 180908820-180908820 on Build GRCh38
rs777395864 Chr.5: 180908738-180908738 on Build GRCh38
rs77209167 Chr.5: 180908923-180908923 on Build GRCh38
rs141841020 Chr.5: 180908817-180908817 on Build GRCh38
rs756910889 Chr.5: 180908892-180908892 on Build GRCh38
rs769176293 Chr.5: 180908753-180908753 on Build GRCh38
rs776901568 Chr.5: 180908754-180908754 on Build GRCh38
rs763180036 Chr.5: 180908776-180908776 on Build GRCh38

View Full Product Details