Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53210207-53210432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT2
Entrez Gene ID
1376
Gene Name
carnitine palmitoyltransferase 2
Gene Aliases
CPT1, CPTASE, IIAE4
UniGene
Hs.713535
Gene Chromosome Location
Chr.1: 53196429-53214197 on Build GRCh38
Transcripts
NM_000098.2 XM_005270484.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4674502 c.686C>A p.P229H Chr.1: 53210360-53210360 on Build GRCh38
7244633 c.744A>G p.K248K Chr.1: 53210418-53210418 on Build GRCh38
5569513 c.707C>G p.T236S Chr.1: 53210381-53210381 on Build GRCh38
5498511 c.550G>A p.D184N Chr.1: 53210224-53210224 on Build GRCh38
5451346 c.745G>T p.G249* Chr.1: 53210419-53210419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747845465 Chr.1: 53210308-53210308 on Build GRCh38
rs753270905 Chr.1: 53210351-53210351 on Build GRCh38
rs973697337 Chr.1: 53210310-53210310 on Build GRCh38
rs760981841 Chr.1: 53210265-53210266 on Build GRCh38
rs111896041 Chr.1: 53210399-53210399 on Build GRCh38
rs766575997 Chr.1: 53210243-53210243 on Build GRCh38
rs367691827 Chr.1: 53210329-53210329 on Build GRCh38
rs144805101 Chr.1: 53210372-53210372 on Build GRCh38
rs776833476 Chr.1: 53210341-53210341 on Build GRCh38
rs377737941 Chr.1: 53210230-53210230 on Build GRCh38
rs759423137 Chr.1: 53210391-53210391 on Build GRCh38
rs1044393692 Chr.1: 53210232-53210232 on Build GRCh38
rs201622710 Chr.1: 53210307-53210307 on Build GRCh38
rs886046409 Chr.1: 53210357-53210357 on Build GRCh38
rs373638740 Chr.1: 53210365-53210365 on Build GRCh38
rs796051944 Chr.1: 53210393-53210393 on Build GRCh38
rs778655162 Chr.1: 53210398-53210398 on Build GRCh38
rs762366252 Chr.1: 53210343-53210343 on Build GRCh38
rs760978599 Chr.1: 53210339-53210339 on Build GRCh38
rs920941550 Chr.1: 53210330-53210330 on Build GRCh38
rs794727616 Chr.1: 53210348-53210348 on Build GRCh38
rs140853350 Chr.1: 53210262-53210262 on Build GRCh38
rs867555821 Chr.1: 53210285-53210285 on Build GRCh38
rs752975688 Chr.1: 53210253-53210253 on Build GRCh38
rs760775391 Chr.1: 53210239-53210239 on Build GRCh38
rs754042241 Chr.1: 53210249-53210249 on Build GRCh38
rs74315300 Chr.1: 53210312-53210312 on Build GRCh38
rs796051942 Chr.1: 53210305-53210305 on Build GRCh38
rs1016498296 Chr.1: 53210293-53210293 on Build GRCh38
rs773503132 Chr.1: 53210237-53210237 on Build GRCh38
rs748844820 Chr.1: 53210298-53210298 on Build GRCh38
rs753303896 Chr.1: 53210388-53210388 on Build GRCh38
rs934539178 Chr.1: 53210368-53210368 on Build GRCh38
rs515726174 Chr.1: 53210315-53210315 on Build GRCh38
rs147889730 Chr.1: 53210350-53210350 on Build GRCh38
rs200252755 Chr.1: 53210395-53210395 on Build GRCh38
rs766770153 Chr.1: 53210340-53210340 on Build GRCh38
rs767592132 Chr.1: 53210235-53210235 on Build GRCh38
rs369369333 Chr.1: 53210366-53210366 on Build GRCh38
rs765824169 Chr.1: 53210252-53210252 on Build GRCh38
rs755830520 Chr.1: 53210280-53210280 on Build GRCh38
rs779525393 Chr.1: 53210288-53210288 on Build GRCh38
rs773236568 Chr.1: 53210331-53210331 on Build GRCh38
rs758823353 Chr.1: 53210261-53210261 on Build GRCh38
rs905917368 Chr.1: 53210248-53210248 on Build GRCh38
rs769115843 Chr.1: 53210278-53210279 on Build GRCh38
rs774374525 Chr.1: 53210301-53210301 on Build GRCh38
rs759733220 Chr.1: 53210347-53210347 on Build GRCh38
rs530706050 Chr.1: 53210367-53210367 on Build GRCh38
rs375968699 Chr.1: 53210251-53210251 on Build GRCh38
rs773788921 Chr.1: 53210300-53210300 on Build GRCh38
rs74315298 Chr.1: 53210354-53210354 on Build GRCh38
rs745547578 Chr.1: 53210277-53210277 on Build GRCh38
rs777297837 Chr.1: 53210285-53210285 on Build GRCh38

View Full Product Details