Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49716102-49716375 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117A
Entrez Gene ID
81558
Gene Name
family with sequence similarity 117 member A
Gene Aliases
-
UniGene
Hs.514308
Gene Chromosome Location
Chr.17: 49710325-49764156 on Build GRCh38
Transcripts
NM_030802.3 XM_017025182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251710 c.937C>T p.L313F Chr.17: 49716289-49716289 on Build GRCh38
5554157 c.1008C>T p.F336F Chr.17: 49716218-49716218 on Build GRCh38
1479706 c.917C>T p.S306F Chr.17: 49716309-49716309 on Build GRCh38
1750123 c.1061C>T p.T354M Chr.17: 49716165-49716165 on Build GRCh38
6406860 c.974C>T p.A325V Chr.17: 49716252-49716252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919454586 Chr.17: 49716274-49716274 on Build GRCh38
rs750574327 Chr.17: 49716305-49716305 on Build GRCh38
rs764189202 Chr.17: 49716140-49716140 on Build GRCh38
rs758646392 Chr.17: 49716311-49716311 on Build GRCh38
rs746156669 Chr.17: 49716184-49716184 on Build GRCh38
rs767435480 Chr.17: 49716128-49716128 on Build GRCh38
rs986107693 Chr.17: 49716209-49716209 on Build GRCh38
rs751899031 Chr.17: 49716318-49716318 on Build GRCh38
rs748714664 Chr.17: 49716225-49716225 on Build GRCh38
rs756152412 Chr.17: 49716137-49716137 on Build GRCh38
rs753844592 Chr.17: 49716157-49716157 on Build GRCh38
rs773819517 Chr.17: 49716232-49716232 on Build GRCh38
rs771953442 Chr.17: 49716245-49716245 on Build GRCh38
rs555059061 Chr.17: 49716295-49716295 on Build GRCh38
rs1022213525 Chr.17: 49716165-49716165 on Build GRCh38
rs770540422 Chr.17: 49716231-49716231 on Build GRCh38
rs879461177 Chr.17: 49716270-49716270 on Build GRCh38
rs376045336 Chr.17: 49716257-49716257 on Build GRCh38
rs781544002 Chr.17: 49716332-49716332 on Build GRCh38
rs780397368 Chr.17: 49716207-49716207 on Build GRCh38
rs769173461 Chr.17: 49716213-49716213 on Build GRCh38
rs753882843 Chr.17: 49716285-49716285 on Build GRCh38
rs757401112 Chr.17: 49716167-49716167 on Build GRCh38
rs568571185 Chr.17: 49716317-49716317 on Build GRCh38
rs114872025 Chr.17: 49716183-49716183 on Build GRCh38
rs755409557 Chr.17: 49716328-49716328 on Build GRCh38
rs758701786 Chr.17: 49716186-49716186 on Build GRCh38
rs151068437 Chr.17: 49716341-49716341 on Build GRCh38
rs752617500 Chr.17: 49716129-49716129 on Build GRCh38
rs999213312 Chr.17: 49716143-49716143 on Build GRCh38
rs142144090 Chr.17: 49716211-49716211 on Build GRCh38
rs151199188 Chr.17: 49716283-49716283 on Build GRCh38
rs966329084 Chr.17: 49716127-49716127 on Build GRCh38
rs759268495 Chr.17: 49716241-49716241 on Build GRCh38
rs560116483 Chr.17: 49716214-49716214 on Build GRCh38

View Full Product Details