Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48469291-48469540 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH2
Entrez Gene ID
9266
Gene Name
cytohesin 2
Gene Aliases
ARNO, CTS18, CTS18.1, PSCD2, PSCD2L, SEC7L, Sec7p-L, Sec7p-like
UniGene
Hs.144011
Gene Chromosome Location
Chr.19: 48469208-48482314 on Build GRCh38
Transcripts
NM_004228.6 NM_017457.5

Gene Symbol
LOC105372430
Entrez Gene ID
105372430
Gene Name
uncharacterized LOC105372430
Gene Aliases
-
UniGene
Hs.736972
Gene Chromosome Location
Chr.19: 48465471-48469507 on Build GRCh38
Transcripts
NM_004228.6 NM_017457.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6249711 c.19+4G>T p.? Chr.19: 48469530-48469530 on Build GRCh38
6249712 c.19+4G>T p.? Chr.19: 48469530-48469530 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs8106937 Chr.19: 48469483-48469483 on Build GRCh38
rs11292058 Chr.19: 48469357-48469357 on Build GRCh38
rs550245290 Chr.19: 48469486-48469486 on Build GRCh38
rs1025387688 Chr.19: 48469321-48469321 on Build GRCh38
rs373320119 Chr.19: 48469487-48469487 on Build GRCh38
rs959425365 Chr.19: 48469425-48469425 on Build GRCh38
rs560233946 Chr.19: 48469393-48469393 on Build GRCh38
rs937282050 Chr.19: 48469511-48469511 on Build GRCh38
rs1024548570 Chr.19: 48469434-48469434 on Build GRCh38
rs761479391 Chr.19: 48469492-48469492 on Build GRCh38
rs544227705 Chr.19: 48469351-48469351 on Build GRCh38
rs752626109 Chr.19: 48469451-48469451 on Build GRCh38
rs574502997 Chr.19: 48469386-48469386 on Build GRCh38
rs541875542 Chr.19: 48469392-48469392 on Build GRCh38
rs1033655074 Chr.19: 48469396-48469396 on Build GRCh38
rs964347474 Chr.19: 48469358-48469359 on Build GRCh38
rs372002789 Chr.19: 48469376-48469376 on Build GRCh38
rs868169177 Chr.19: 48469431-48469431 on Build GRCh38
rs1049297127 Chr.19: 48469472-48469472 on Build GRCh38
rs546363096 Chr.19: 48469450-48469450 on Build GRCh38
rs1017588607 Chr.19: 48469357-48469357 on Build GRCh38
rs746567865 Chr.19: 48469485-48469485 on Build GRCh38
rs189344954 Chr.19: 48469363-48469363 on Build GRCh38
rs564552690 Chr.19: 48469462-48469462 on Build GRCh38
rs796935769 Chr.19: 48469358-48469358 on Build GRCh38
rs527846893 Chr.19: 48469442-48469442 on Build GRCh38

View Full Product Details