Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65074189-65074462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP68
Entrez Gene ID
23177
Gene Name
centrosomal protein 68
Gene Aliases
KIAA0582
UniGene
Hs.709257
Gene Chromosome Location
Chr.2: 65056361-65087008 on Build GRCh38
Transcripts
NM_001319100.1 NM_001319101.1 NM_015147.2 NR_134966.1 XM_005264219.3 XM_017003633.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6276479 c.1995G>T p.L665L Chr.2: 65074392-65074392 on Build GRCh38
307732 c.1964G>A p.R655K Chr.2: 65074361-65074361 on Build GRCh38
405317 c.1966T>G p.S656A Chr.2: 65074363-65074363 on Build GRCh38
3743861 c.1885A>T p.T629S Chr.2: 65074282-65074282 on Build GRCh38
4475645 c.2004C>T p.Y668Y Chr.2: 65074401-65074401 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572089458 Chr.2: 65074414-65074414 on Build GRCh38
rs779138543 Chr.2: 65074402-65074402 on Build GRCh38
rs137938370 Chr.2: 65074367-65074367 on Build GRCh38
rs376614865 Chr.2: 65074320-65074320 on Build GRCh38
rs79568756 Chr.2: 65074413-65074413 on Build GRCh38
rs763725759 Chr.2: 65074308-65074308 on Build GRCh38
rs373587824 Chr.2: 65074277-65074277 on Build GRCh38
rs747266373 Chr.2: 65074339-65074340 on Build GRCh38
rs373794271 Chr.2: 65074243-65074243 on Build GRCh38
rs749429654 Chr.2: 65074240-65074240 on Build GRCh38
rs1009226308 Chr.2: 65074373-65074373 on Build GRCh38
rs760078030 Chr.2: 65074347-65074347 on Build GRCh38
rs772357679 Chr.2: 65074411-65074411 on Build GRCh38
rs200007619 Chr.2: 65074396-65074396 on Build GRCh38
rs976523774 Chr.2: 65074417-65074417 on Build GRCh38
rs756558196 Chr.2: 65074352-65074352 on Build GRCh38
rs1048465640 Chr.2: 65074217-65074217 on Build GRCh38
rs762895922 Chr.2: 65074418-65074418 on Build GRCh38
rs761083973 Chr.2: 65074236-65074236 on Build GRCh38
rs749364217 Chr.2: 65074382-65074382 on Build GRCh38
rs910007885 Chr.2: 65074224-65074224 on Build GRCh38
rs901401430 Chr.2: 65074274-65074274 on Build GRCh38
rs932290116 Chr.2: 65074219-65074219 on Build GRCh38
rs201117405 Chr.2: 65074350-65074350 on Build GRCh38
rs893596089 Chr.2: 65074355-65074355 on Build GRCh38
rs762520281 Chr.2: 65074299-65074299 on Build GRCh38
rs770564543 Chr.2: 65074255-65074255 on Build GRCh38
rs780220102 Chr.2: 65074276-65074276 on Build GRCh38
rs753879555 Chr.2: 65074322-65074322 on Build GRCh38
rs6713125 Chr.2: 65074284-65074284 on Build GRCh38
rs569855265 Chr.2: 65074276-65074276 on Build GRCh38
rs1036870988 Chr.2: 65074243-65074244 on Build GRCh38
rs114558502 Chr.2: 65074314-65074314 on Build GRCh38
rs751112426 Chr.2: 65074429-65074429 on Build GRCh38
rs768083390 Chr.2: 65074403-65074403 on Build GRCh38
rs768995032 Chr.2: 65074242-65074242 on Build GRCh38
rs765205235 Chr.2: 65074278-65074278 on Build GRCh38

View Full Product Details