Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57488861-57489086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC43A1
Entrez Gene ID
8501
Gene Name
solute carrier family 43 member 1
Gene Aliases
LAT3, PB39, POV1, R00504
UniGene
Hs.591952
Gene Chromosome Location
Chr.11: 57484531-57515954 on Build GRCh38
Transcripts
NM_001198810.1 NM_003627.5 XM_005274358.4 XM_011545320.2 XM_011545321.2 XM_011545322.1 XM_017018451.1 XM_017018452.1 XM_017018453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7200711 c.1372T>C p.F458L Chr.11: 57488953-57488953 on Build GRCh38
1492575 c.1336-2A>T p.? Chr.11: 57488991-57488991 on Build GRCh38
6228585 c.1366C>T p.R456* Chr.11: 57488959-57488959 on Build GRCh38
3869554 c.1374C>T p.F458F Chr.11: 57488951-57488951 on Build GRCh38
7093156 c.1364T>C p.V455A Chr.11: 57488961-57488961 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778089122 Chr.11: 57488902-57488902 on Build GRCh38
rs891956157 Chr.11: 57488939-57488939 on Build GRCh38
rs749406908 Chr.11: 57488904-57488904 on Build GRCh38
rs568693252 Chr.11: 57489038-57489038 on Build GRCh38
rs201763475 Chr.11: 57488920-57488920 on Build GRCh38
rs761325929 Chr.11: 57488937-57488937 on Build GRCh38
rs150971228 Chr.11: 57488959-57488959 on Build GRCh38
rs766855806 Chr.11: 57489026-57489027 on Build GRCh38
rs36069501 Chr.11: 57489057-57489057 on Build GRCh38
rs200502867 Chr.11: 57488931-57488931 on Build GRCh38
rs1005179575 Chr.11: 57489059-57489059 on Build GRCh38
rs772912807 Chr.11: 57488955-57488955 on Build GRCh38
rs564460933 Chr.11: 57488965-57488965 on Build GRCh38
rs775335116 Chr.11: 57488926-57488926 on Build GRCh38
rs753123785 Chr.11: 57488970-57488970 on Build GRCh38
rs372673734 Chr.11: 57489016-57489016 on Build GRCh38
rs754160476 Chr.11: 57489038-57489050 on Build GRCh38
rs1051982161 Chr.11: 57488923-57488923 on Build GRCh38
rs757359121 Chr.11: 57488918-57488918 on Build GRCh38
rs114429407 Chr.11: 57488971-57488971 on Build GRCh38
rs780100674 Chr.11: 57489022-57489022 on Build GRCh38
rs776365398 Chr.11: 57488934-57488934 on Build GRCh38
rs772035105 Chr.11: 57488925-57488925 on Build GRCh38
rs1042824591 Chr.11: 57488972-57488972 on Build GRCh38
rs764712852 Chr.11: 57488945-57488945 on Build GRCh38
rs759754295 Chr.11: 57489058-57489058 on Build GRCh38
rs995865279 Chr.11: 57489052-57489052 on Build GRCh38
rs58975956 Chr.11: 57489046-57489046 on Build GRCh38
rs899360224 Chr.11: 57489009-57489009 on Build GRCh38
rs761147422 Chr.11: 57488888-57488888 on Build GRCh38
rs1016871510 Chr.11: 57489002-57489003 on Build GRCh38
rs760571627 Chr.11: 57488929-57488929 on Build GRCh38
rs764678946 Chr.11: 57488974-57488974 on Build GRCh38
rs757481985 Chr.11: 57488991-57488991 on Build GRCh38
rs376477109 Chr.11: 57488901-57488901 on Build GRCh38
rs961455114 Chr.11: 57488893-57488893 on Build GRCh38
rs375196672 Chr.11: 57488997-57488997 on Build GRCh38
rs547081389 Chr.11: 57489007-57489007 on Build GRCh38
rs754387663 Chr.11: 57488977-57488977 on Build GRCh38
rs768669485 Chr.11: 57488933-57488933 on Build GRCh38

View Full Product Details