Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 146292844-146293115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC10A7
Entrez Gene ID
84068
Gene Name
solute carrier family 10 member 7
Gene Aliases
C4orf13, P7
UniGene
Hs.659209
Gene Chromosome Location
Chr.4: 146253981-146522351 on Build GRCh38
Transcripts
NM_001029998.5 NM_001300842.2 NM_001317816.1 NR_133924.1 XM_011532311.2 XM_011532313.2 XM_011532314.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5709096 c.752T>C p.L251S Chr.4: 146292950-146292950 on Build GRCh38
4785847 c.732C>T p.I244I Chr.4: 146292970-146292970 on Build GRCh38
5865842 c.728C>T p.S243F Chr.4: 146292974-146292974 on Build GRCh38
1427405 c.732C>T p.I244I Chr.4: 146292970-146292970 on Build GRCh38
5865843 c.728C>T p.S243F Chr.4: 146292974-146292974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780647325 Chr.4: 146293033-146293044 on Build GRCh38
rs754697850 Chr.4: 146292953-146292953 on Build GRCh38
rs766749886 Chr.4: 146293024-146293024 on Build GRCh38
rs892182157 Chr.4: 146292928-146292928 on Build GRCh38
rs777925649 Chr.4: 146292896-146292896 on Build GRCh38
rs1021894216 Chr.4: 146292947-146292947 on Build GRCh38
rs778223908 Chr.4: 146292907-146292907 on Build GRCh38
rs113764915 Chr.4: 146292956-146292956 on Build GRCh38
rs199983559 Chr.4: 146292904-146292904 on Build GRCh38
rs748508836 Chr.4: 146293023-146293023 on Build GRCh38
rs758482849 Chr.4: 146292884-146292884 on Build GRCh38
rs189095643 Chr.4: 146293036-146293036 on Build GRCh38
rs769001734 Chr.4: 146292967-146292967 on Build GRCh38
rs530503649 Chr.4: 146292894-146292894 on Build GRCh38
rs369056739 Chr.4: 146292972-146292972 on Build GRCh38
rs976242172 Chr.4: 146293056-146293056 on Build GRCh38
rs750672096 Chr.4: 146292881-146292881 on Build GRCh38
rs769921140 Chr.4: 146292995-146292995 on Build GRCh38
rs771572173 Chr.4: 146292929-146292929 on Build GRCh38
rs1025216494 Chr.4: 146292994-146292994 on Build GRCh38
rs773117913 Chr.4: 146292996-146292996 on Build GRCh38
rs549093817 Chr.4: 146293079-146293079 on Build GRCh38
rs775061906 Chr.4: 146292935-146292935 on Build GRCh38
rs777178582 Chr.4: 146292969-146292969 on Build GRCh38
rs76035000 Chr.4: 146293042-146293042 on Build GRCh38
rs555266661 Chr.4: 146292987-146292987 on Build GRCh38
rs12711434 Chr.4: 146292877-146292877 on Build GRCh38
rs903511317 Chr.4: 146292951-146292951 on Build GRCh38
rs377005227 Chr.4: 146292932-146292932 on Build GRCh38
rs769216178 Chr.4: 146292886-146292886 on Build GRCh38
rs546843671 Chr.4: 146293039-146293039 on Build GRCh38
rs763066205 Chr.4: 146293020-146293020 on Build GRCh38
rs753391766 Chr.4: 146292917-146292918 on Build GRCh38
rs551205022 Chr.4: 146292903-146292903 on Build GRCh38
rs1042830778 Chr.4: 146292937-146292937 on Build GRCh38
rs752589177 Chr.4: 146292888-146292888 on Build GRCh38
rs745569875 Chr.4: 146292912-146292912 on Build GRCh38
rs745380882 Chr.4: 146292880-146292880 on Build GRCh38
rs147545268 Chr.4: 146293060-146293060 on Build GRCh38

View Full Product Details