Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55762364-55762625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFPL4A
Entrez Gene ID
342931
Gene Name
ret finger protein like 4A
Gene Aliases
RFPL4, RNF210
UniGene
Hs.631553
Gene Chromosome Location
Chr.19: 55757298-55763388 on Build GRCh38
Transcripts
NM_001145014.1 XM_011526915.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2969386 c.301G>A p.D101N Chr.19: 55762612-55762612 on Build GRCh38
2969385 c.300C>T p.F100F Chr.19: 55762611-55762611 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573223365 Chr.19: 55762511-55762511 on Build GRCh38
rs543736713 Chr.19: 55762576-55762576 on Build GRCh38
rs768754237 Chr.19: 55762602-55762602 on Build GRCh38
rs377529210 Chr.19: 55762487-55762487 on Build GRCh38
rs553341622 Chr.19: 55762417-55762417 on Build GRCh38
rs750097514 Chr.19: 55762556-55762556 on Build GRCh38
rs2045865 Chr.19: 55762536-55762536 on Build GRCh38
rs779642278 Chr.19: 55762562-55762562 on Build GRCh38
rs78193304 Chr.19: 55762412-55762412 on Build GRCh38
rs760871241 Chr.19: 55762605-55762605 on Build GRCh38
rs2045866 Chr.19: 55762538-55762538 on Build GRCh38
rs577951269 Chr.19: 55762549-55762549 on Build GRCh38
rs200686834 Chr.19: 55762502-55762502 on Build GRCh38
rs201406017 Chr.19: 55762504-55762504 on Build GRCh38
rs755669740 Chr.19: 55762558-55762558 on Build GRCh38
rs778974887 Chr.19: 55762607-55762607 on Build GRCh38

View Full Product Details