Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 23792832-23793077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYPLA2
Entrez Gene ID
11313
Gene Name
lysophospholipase II
Gene Aliases
APT-2, APT2, DJ886K2.4
UniGene
Hs.533479
Gene Chromosome Location
Chr.1: 23790789-23795539 on Build GRCh38
Transcripts
NM_007260.2 XM_005245728.4 XM_017000165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3685947 c.83delT p.L30fs*35 Chr.1: 23793012-23793012 on Build GRCh38
6705334 c.105C>T p.D35D Chr.1: 23793034-23793034 on Build GRCh38
2049816 c.83delT p.L30fs*35 Chr.1: 23793012-23793012 on Build GRCh38
6705335 c.105C>T p.D35D Chr.1: 23793034-23793034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756191714 Chr.1: 23792976-23792976 on Build GRCh38
rs748075123 Chr.1: 23793037-23793037 on Build GRCh38
rs766716526 Chr.1: 23792969-23792969 on Build GRCh38
rs777462448 Chr.1: 23793054-23793055 on Build GRCh38
rs904531463 Chr.1: 23792981-23792981 on Build GRCh38
rs777767043 Chr.1: 23792977-23792977 on Build GRCh38
rs371527090 Chr.1: 23792986-23792986 on Build GRCh38
rs372601977 Chr.1: 23792963-23792963 on Build GRCh38
rs992745530 Chr.1: 23792971-23792971 on Build GRCh38
rs2232979 Chr.1: 23792893-23792893 on Build GRCh38
rs746815475 Chr.1: 23792999-23792999 on Build GRCh38
rs1034139289 Chr.1: 23792935-23792935 on Build GRCh38
rs550681434 Chr.1: 23792882-23792882 on Build GRCh38
rs563036143 Chr.1: 23792883-23792883 on Build GRCh38
rs368623413 Chr.1: 23792958-23792958 on Build GRCh38
rs752344350 Chr.1: 23792968-23792969 on Build GRCh38
rs374787618 Chr.1: 23793005-23793005 on Build GRCh38
rs968526176 Chr.1: 23792925-23792925 on Build GRCh38
rs779754947 Chr.1: 23792994-23792994 on Build GRCh38
rs537012768 Chr.1: 23792987-23792987 on Build GRCh38
rs2232980 Chr.1: 23792970-23792970 on Build GRCh38
rs369248220 Chr.1: 23793050-23793050 on Build GRCh38
rs768494389 Chr.1: 23793001-23793001 on Build GRCh38
rs2232981 Chr.1: 23792983-23792983 on Build GRCh38
rs772617928 Chr.1: 23793053-23793053 on Build GRCh38
rs1020302800 Chr.1: 23792899-23792899 on Build GRCh38
rs755891364 Chr.1: 23793050-23793051 on Build GRCh38
rs978532877 Chr.1: 23792930-23792930 on Build GRCh38
rs768125088 Chr.1: 23792973-23792973 on Build GRCh38
rs762601559 Chr.1: 23793058-23793058 on Build GRCh38
rs112266814 Chr.1: 23792974-23792974 on Build GRCh38

View Full Product Details