Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 20937691-20937964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELL1
Entrez Gene ID
4745
Gene Name
neural EGFL like 1
Gene Aliases
IDH3GL, NRP1
UniGene
Hs.657172
Gene Chromosome Location
Chr.11: 20669551-21575686 on Build GRCh38
Transcripts
NM_001288713.1 NM_001288714.1 NM_006157.4 NM_201551.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1675893 c.1030G>A p.E344K Chr.11: 20937818-20937818 on Build GRCh38
1258988 c.1023T>C p.V341V Chr.11: 20937811-20937811 on Build GRCh38
1258990 c.1000A>C p.K334Q Chr.11: 20937788-20937788 on Build GRCh38
1703718 c.1028C>G p.A343G Chr.11: 20937816-20937816 on Build GRCh38
1703717 c.1012G>A p.G338R Chr.11: 20937800-20937800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528105353 Chr.11: 20937829-20937829 on Build GRCh38
rs756238269 Chr.11: 20937889-20937889 on Build GRCh38
rs781489940 Chr.11: 20937767-20937767 on Build GRCh38
rs192271295 Chr.11: 20937763-20937763 on Build GRCh38
rs1880087 Chr.11: 20937883-20937883 on Build GRCh38
rs745848313 Chr.11: 20937825-20937825 on Build GRCh38
rs551356049 Chr.11: 20937846-20937846 on Build GRCh38
rs373855331 Chr.11: 20937756-20937756 on Build GRCh38
rs748111769 Chr.11: 20937772-20937772 on Build GRCh38
rs993931465 Chr.11: 20937799-20937799 on Build GRCh38
rs372576607 Chr.11: 20937735-20937735 on Build GRCh38
rs750381072 Chr.11: 20937903-20937903 on Build GRCh38
rs1880086 Chr.11: 20937936-20937936 on Build GRCh38
rs752984709 Chr.11: 20937884-20937884 on Build GRCh38
rs964087866 Chr.11: 20937860-20937860 on Build GRCh38
rs1005739389 Chr.11: 20937813-20937813 on Build GRCh38
rs778039121 Chr.11: 20937900-20937900 on Build GRCh38
rs183315828 Chr.11: 20937924-20937924 on Build GRCh38
rs188049726 Chr.11: 20937927-20937927 on Build GRCh38
rs968504384 Chr.11: 20937917-20937917 on Build GRCh38
rs8176786 Chr.11: 20937848-20937848 on Build GRCh38
rs751814170 Chr.11: 20937870-20937870 on Build GRCh38
rs1050593950 Chr.11: 20937750-20937750 on Build GRCh38
rs771961538 Chr.11: 20937828-20937828 on Build GRCh38
rs923547533 Chr.11: 20937726-20937726 on Build GRCh38
rs778944309 Chr.11: 20937798-20937798 on Build GRCh38
rs765663394 Chr.11: 20937742-20937742 on Build GRCh38
rs994346839 Chr.11: 20937758-20937758 on Build GRCh38
rs776784394 Chr.11: 20937816-20937816 on Build GRCh38
rs8176791 Chr.11: 20937919-20937919 on Build GRCh38
rs761182277 Chr.11: 20937849-20937849 on Build GRCh38
rs780060086 Chr.11: 20937841-20937841 on Build GRCh38
rs749747863 Chr.11: 20937893-20937893 on Build GRCh38
rs377277061 Chr.11: 20937748-20937748 on Build GRCh38
rs367598660 Chr.11: 20937819-20937819 on Build GRCh38
rs371903884 Chr.11: 20937857-20937857 on Build GRCh38
rs766370630 Chr.11: 20937858-20937858 on Build GRCh38
rs756250761 Chr.11: 20937793-20937793 on Build GRCh38
rs761398046 Chr.11: 20937737-20937737 on Build GRCh38
rs759951255 Chr.11: 20937880-20937880 on Build GRCh38
rs766717707 Chr.11: 20937869-20937869 on Build GRCh38
rs896076196 Chr.11: 20937751-20937751 on Build GRCh38

View Full Product Details