Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19922255-19922514 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G2E
Entrez Gene ID
30814
Gene Name
phospholipase A2 group IIE
Gene Aliases
GIIE sPLA2, sPLA2-IIE
UniGene
Hs.272372
Gene Chromosome Location
Chr.1: 19920307-19923617 on Build GRCh38
Transcripts
NM_014589.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902343 c.214C>A p.L72M Chr.1: 19922370-19922370 on Build GRCh38
902344 c.211C>A p.R71S Chr.1: 19922373-19922373 on Build GRCh38
3481717 c.209G>A p.G70E Chr.1: 19922375-19922375 on Build GRCh38
4925628 c.226G>A p.G76S Chr.1: 19922358-19922358 on Build GRCh38
393440 c.268G>C p.E90Q Chr.1: 19922316-19922316 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61729971 Chr.1: 19922372-19922372 on Build GRCh38
rs771769821 Chr.1: 19922320-19922320 on Build GRCh38
rs559232898 Chr.1: 19922313-19922313 on Build GRCh38
rs200544567 Chr.1: 19922441-19922441 on Build GRCh38
rs761679302 Chr.1: 19922417-19922417 on Build GRCh38
rs150118869 Chr.1: 19922354-19922354 on Build GRCh38
rs145027583 Chr.1: 19922302-19922302 on Build GRCh38
rs546930480 Chr.1: 19922475-19922475 on Build GRCh38
rs372491665 Chr.1: 19922377-19922377 on Build GRCh38
rs994176586 Chr.1: 19922371-19922371 on Build GRCh38
rs780494033 Chr.1: 19922373-19922373 on Build GRCh38
rs781437830 Chr.1: 19922378-19922378 on Build GRCh38
rs768315780 Chr.1: 19922413-19922413 on Build GRCh38
rs528463608 Chr.1: 19922470-19922470 on Build GRCh38
rs775005614 Chr.1: 19922399-19922399 on Build GRCh38
rs1044608406 Chr.1: 19922478-19922478 on Build GRCh38
rs763808244 Chr.1: 19922336-19922336 on Build GRCh38
rs778239539 Chr.1: 19922388-19922388 on Build GRCh38
rs201254783 Chr.1: 19922308-19922308 on Build GRCh38
rs973551247 Chr.1: 19922472-19922472 on Build GRCh38
rs370199149 Chr.1: 19922356-19922356 on Build GRCh38
rs535636403 Chr.1: 19922391-19922391 on Build GRCh38
rs751738554 Chr.1: 19922376-19922376 on Build GRCh38
rs140744541 Chr.1: 19922385-19922385 on Build GRCh38
rs949842069 Chr.1: 19922469-19922469 on Build GRCh38
rs760268843 Chr.1: 19922410-19922410 on Build GRCh38
rs916971981 Chr.1: 19922455-19922455 on Build GRCh38
rs760306746 Chr.1: 19922331-19922331 on Build GRCh38
rs765327223 Chr.1: 19922422-19922422 on Build GRCh38
rs1010448715 Chr.1: 19922389-19922389 on Build GRCh38
rs745903576 Chr.1: 19922285-19922285 on Build GRCh38
rs367824617 Chr.1: 19922316-19922316 on Build GRCh38
rs988437369 Chr.1: 19922442-19922442 on Build GRCh38
rs778839822 Chr.1: 19922315-19922315 on Build GRCh38
rs184234254 Chr.1: 19922457-19922457 on Build GRCh38
rs773902416 Chr.1: 19922350-19922350 on Build GRCh38
rs533011527 Chr.1: 19922342-19922342 on Build GRCh38
rs372534405 Chr.1: 19922416-19922416 on Build GRCh38
rs766582490 Chr.1: 19922446-19922446 on Build GRCh38
rs780404061 Chr.1: 19922293-19922293 on Build GRCh38
rs775119539 Chr.1: 19922322-19922322 on Build GRCh38
rs374893489 Chr.1: 19922299-19922299 on Build GRCh38
rs74059111 Chr.1: 19922434-19922434 on Build GRCh38
rs369294108 Chr.1: 19922395-19922395 on Build GRCh38
rs770331034 Chr.1: 19922386-19922386 on Build GRCh38
rs758734845 Chr.1: 19922289-19922289 on Build GRCh38

View Full Product Details