Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 154049806-154050053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFAP3
Entrez Gene ID
4238
Gene Name
microfibrillar associated protein 3
Gene Aliases
-
UniGene
Hs.432818
Gene Chromosome Location
Chr.5: 154038959-154057454 on Build GRCh38
Transcripts
NM_001135037.1 NM_001242336.1 NM_005927.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1619880 c.156A>T p.A52A Chr.5: 154049878-154049878 on Build GRCh38
3722516 c.177T>C p.D59D Chr.5: 154049899-154049899 on Build GRCh38
6711805 c.205G>A p.V69I Chr.5: 154049927-154049927 on Build GRCh38
6711804 c.178G>A p.V60I Chr.5: 154049900-154049900 on Build GRCh38
4701865 c.116C>A p.S39Y Chr.5: 154049838-154049838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754125012 Chr.5: 154049861-154049861 on Build GRCh38
rs139479394 Chr.5: 154049926-154049926 on Build GRCh38
rs761733582 Chr.5: 154049831-154049831 on Build GRCh38
rs779082087 Chr.5: 154049863-154049863 on Build GRCh38
rs769098960 Chr.5: 154049994-154049994 on Build GRCh38
rs371882857 Chr.5: 154049931-154049931 on Build GRCh38
rs543568426 Chr.5: 154049841-154049841 on Build GRCh38
rs753192058 Chr.5: 154049857-154049857 on Build GRCh38
rs776089498 Chr.5: 154049923-154049923 on Build GRCh38
rs774055709 Chr.5: 154049843-154049843 on Build GRCh38
rs754014315 Chr.5: 154049961-154049961 on Build GRCh38
rs759103854 Chr.5: 154049844-154049844 on Build GRCh38
rs690592 Chr.5: 154049899-154049899 on Build GRCh38
rs749104208 Chr.5: 154049952-154049952 on Build GRCh38
rs925185142 Chr.5: 154049891-154049891 on Build GRCh38
rs756056668 Chr.5: 154049947-154049947 on Build GRCh38
rs747977002 Chr.5: 154049890-154049890 on Build GRCh38
rs757600557 Chr.5: 154049928-154049928 on Build GRCh38
rs140399898 Chr.5: 154049892-154049892 on Build GRCh38
rs750576275 Chr.5: 154049933-154049933 on Build GRCh38
rs766100038 Chr.5: 154049842-154049842 on Build GRCh38
rs142652576 Chr.5: 154049927-154049927 on Build GRCh38
rs1030098107 Chr.5: 154049845-154049845 on Build GRCh38
rs758486016 Chr.5: 154049934-154049934 on Build GRCh38
rs774906082 Chr.5: 154049987-154049987 on Build GRCh38
rs369113358 Chr.5: 154049889-154049889 on Build GRCh38
rs553972755 Chr.5: 154049859-154049859 on Build GRCh38
rs770767723 Chr.5: 154049903-154049903 on Build GRCh38
rs977978151 Chr.5: 154049876-154049876 on Build GRCh38
rs774143509 Chr.5: 154049906-154049906 on Build GRCh38
rs377593609 Chr.5: 154049913-154049913 on Build GRCh38
rs545780539 Chr.5: 154049870-154049870 on Build GRCh38
rs140803939 Chr.5: 154049973-154049973 on Build GRCh38
rs748271641 Chr.5: 154049879-154049879 on Build GRCh38
rs756518710 Chr.5: 154049858-154049858 on Build GRCh38
rs151011744 Chr.5: 154049944-154049944 on Build GRCh38
rs918702979 Chr.5: 154049914-154049914 on Build GRCh38
rs768206640 Chr.5: 154049856-154049856 on Build GRCh38
rs770784204 Chr.5: 154049957-154049957 on Build GRCh38
rs780057486 Chr.5: 154049942-154049942 on Build GRCh38
rs543278918 Chr.5: 154049900-154049900 on Build GRCh38
rs769407854 Chr.5: 154049832-154049832 on Build GRCh38
rs746001365 Chr.5: 154049869-154049869 on Build GRCh38
rs765506224 Chr.5: 154049932-154049932 on Build GRCh38
rs149717807 Chr.5: 154050017-154050017 on Build GRCh38
rs766837435 Chr.5: 154049966-154049966 on Build GRCh38
rs777755246 Chr.5: 154049949-154049949 on Build GRCh38
rs771742684 Chr.5: 154049983-154049983 on Build GRCh38
rs559167658 Chr.5: 154049958-154049958 on Build GRCh38
rs139144281 Chr.5: 154049907-154049907 on Build GRCh38
rs760206793 Chr.5: 154049990-154049990 on Build GRCh38
rs772930272 Chr.5: 154049838-154049838 on Build GRCh38
rs77520992 Chr.5: 154049946-154049946 on Build GRCh38
rs572150652 Chr.5: 154049862-154049862 on Build GRCh38

View Full Product Details