Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10252504-10252735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL4
Entrez Gene ID
51073
Gene Name
mitochondrial ribosomal protein L4
Gene Aliases
CGI-28, L4mt
UniGene
Hs.279652
Gene Chromosome Location
Chr.19: 10251964-10260060 on Build GRCh38
Transcripts
NM_015956.2 NM_146387.1 NM_146388.1 XM_011528045.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6715362 c.222G>T p.Q74H Chr.19: 10252648-10252648 on Build GRCh38
3726978 c.250C>T p.P84S Chr.19: 10252676-10252676 on Build GRCh38
3932859 c.223G>C p.E75Q Chr.19: 10252649-10252649 on Build GRCh38
1480510 c.198C>T p.V66V Chr.19: 10252624-10252624 on Build GRCh38
1563982 c.178C>T p.R60C Chr.19: 10252604-10252604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769993833 Chr.19: 10252683-10252683 on Build GRCh38
rs11552124 Chr.19: 10252668-10252668 on Build GRCh38
rs980169182 Chr.19: 10252693-10252693 on Build GRCh38
rs749098137 Chr.19: 10252652-10252652 on Build GRCh38
rs769734581 Chr.19: 10252583-10252583 on Build GRCh38
rs770900160 Chr.19: 10252657-10252657 on Build GRCh38
rs759648717 Chr.19: 10252694-10252694 on Build GRCh38
rs141266080 Chr.19: 10252596-10252596 on Build GRCh38
rs775168694 Chr.19: 10252605-10252605 on Build GRCh38
rs890250247 Chr.19: 10252593-10252593 on Build GRCh38
rs775080509 Chr.19: 10252673-10252673 on Build GRCh38
rs766888518 Chr.19: 10252689-10252689 on Build GRCh38
rs374374421 Chr.19: 10252669-10252669 on Build GRCh38
rs896200568 Chr.19: 10252564-10252564 on Build GRCh38
rs751673231 Chr.19: 10252535-10252535 on Build GRCh38
rs752999846 Chr.19: 10252649-10252649 on Build GRCh38
rs764292040 Chr.19: 10252701-10252701 on Build GRCh38
rs755910219 Chr.19: 10252651-10252651 on Build GRCh38
rs750828120 Chr.19: 10252617-10252617 on Build GRCh38
rs781517584 Chr.19: 10252647-10252647 on Build GRCh38
rs777536175 Chr.19: 10252589-10252589 on Build GRCh38
rs368731222 Chr.19: 10252631-10252631 on Build GRCh38
rs868508288 Chr.19: 10252537-10252537 on Build GRCh38
rs45520838 Chr.19: 10252604-10252604 on Build GRCh38
rs763269040 Chr.19: 10252687-10252687 on Build GRCh38
rs528482130 Chr.19: 10252614-10252614 on Build GRCh38
rs755249260 Chr.19: 10252538-10252538 on Build GRCh38
rs771589916 Chr.19: 10252670-10252670 on Build GRCh38
rs752913855 Chr.19: 10252696-10252696 on Build GRCh38
rs773611478 Chr.19: 10252686-10252686 on Build GRCh38
rs546944714 Chr.19: 10252642-10252642 on Build GRCh38
rs970267670 Chr.19: 10252655-10252655 on Build GRCh38
rs113197610 Chr.19: 10252575-10252575 on Build GRCh38
rs563534607 Chr.19: 10252526-10252526 on Build GRCh38
rs766851914 Chr.19: 10252629-10252629 on Build GRCh38
rs138990905 Chr.19: 10252634-10252634 on Build GRCh38
rs760324477 Chr.19: 10252680-10252680 on Build GRCh38
rs200328561 Chr.19: 10252700-10252700 on Build GRCh38
rs749257644 Chr.19: 10252595-10252595 on Build GRCh38
rs759110680 Chr.19: 10252601-10252601 on Build GRCh38
rs778753388 Chr.19: 10252659-10252659 on Build GRCh38
rs774742706 Chr.19: 10252692-10252692 on Build GRCh38
rs758369833 Chr.19: 10252527-10252527 on Build GRCh38
rs780186008 Chr.19: 10252530-10252530 on Build GRCh38
rs371590299 Chr.19: 10252610-10252610 on Build GRCh38
rs773901775 Chr.19: 10252600-10252600 on Build GRCh38
rs976156075 Chr.19: 10252703-10252703 on Build GRCh38
rs765706684 Chr.19: 10252524-10252524 on Build GRCh38
rs781607524 Chr.19: 10252561-10252561 on Build GRCh38
rs763527301 Chr.19: 10252620-10252620 on Build GRCh38

View Full Product Details