Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55247198-55247461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6C74
Entrez Gene ID
254783
Gene Name
olfactory receptor family 6 subfamily C member 74
Gene Aliases
-
UniGene
Hs.554556
Gene Chromosome Location
Chr.12: 55244114-55248226 on Build GRCh38
Transcripts
NM_001005490.1 XM_017019126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
313760 c.99C>A p.Y33* Chr.12: 55247386-55247386 on Build GRCh38
5361290 c.21_25delAGCAA p.A8fs*9 Chr.12: 55247308-55247312 on Build GRCh38
5044006 c.34C>G p.L12V Chr.12: 55247321-55247321 on Build GRCh38
5081885 c.107G>T p.S36I Chr.12: 55247394-55247394 on Build GRCh38
5016989 c.13A>G p.T5A Chr.12: 55247300-55247300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752347317 Chr.12: 55247379-55247379 on Build GRCh38
rs113925664 Chr.12: 55247385-55247385 on Build GRCh38
rs761830715 Chr.12: 55247265-55247265 on Build GRCh38
rs775899316 Chr.12: 55247252-55247252 on Build GRCh38
rs755606993 Chr.12: 55247371-55247371 on Build GRCh38
rs754755293 Chr.12: 55247374-55247374 on Build GRCh38
rs375899630 Chr.12: 55247339-55247339 on Build GRCh38
rs11171387 Chr.12: 55247434-55247434 on Build GRCh38
rs946595725 Chr.12: 55247412-55247412 on Build GRCh38
rs985859928 Chr.12: 55247228-55247228 on Build GRCh38
rs767701350 Chr.12: 55247315-55247315 on Build GRCh38
rs370819655 Chr.12: 55247242-55247242 on Build GRCh38
rs7302063 Chr.12: 55247247-55247247 on Build GRCh38
rs974940713 Chr.12: 55247295-55247295 on Build GRCh38
rs749921602 Chr.12: 55247343-55247343 on Build GRCh38
rs915095127 Chr.12: 55247321-55247321 on Build GRCh38
rs897704050 Chr.12: 55247340-55247340 on Build GRCh38
rs749615004 Chr.12: 55247423-55247423 on Build GRCh38
rs746547841 Chr.12: 55247399-55247399 on Build GRCh38
rs777249704 Chr.12: 55247386-55247386 on Build GRCh38
rs753631645 Chr.12: 55247354-55247354 on Build GRCh38
rs780455404 Chr.12: 55247415-55247415 on Build GRCh38
rs769154918 Chr.12: 55247428-55247428 on Build GRCh38
rs746377620 Chr.12: 55247250-55247250 on Build GRCh38
rs887597126 Chr.12: 55247429-55247429 on Build GRCh38
rs778483202 Chr.12: 55247377-55247377 on Build GRCh38
rs750589737 Chr.12: 55247324-55247324 on Build GRCh38
rs777066527 Chr.12: 55247413-55247415 on Build GRCh38
rs368893995 Chr.12: 55247300-55247300 on Build GRCh38
rs7301705 Chr.12: 55247291-55247291 on Build GRCh38
rs761926464 Chr.12: 55247307-55247307 on Build GRCh38
rs770301522 Chr.12: 55247402-55247402 on Build GRCh38
rs762237919 Chr.12: 55247425-55247428 on Build GRCh38
rs777051714 Chr.12: 55247248-55247248 on Build GRCh38

View Full Product Details