Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90268323-90268550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC1
Entrez Gene ID
5700
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
P26S4, S4, p56
UniGene
Hs.356654
Gene Chromosome Location
Chr.14: 90256550-90272625 on Build GRCh38
Transcripts
NM_002802.2 XM_017021470.1 XM_017021471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198974 c.836C>A p.P279Q Chr.14: 90268368-90268368 on Build GRCh38
958612 c.871G>T p.G291W Chr.14: 90268403-90268403 on Build GRCh38
6265147 c.813C>T p.F271F Chr.14: 90268345-90268345 on Build GRCh38
1563061 c.829C>T p.H277Y Chr.14: 90268361-90268361 on Build GRCh38
1629425 c.830A>G p.H277R Chr.14: 90268362-90268362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1031709803 Chr.14: 90268486-90268486 on Build GRCh38
rs996276127 Chr.14: 90268478-90268478 on Build GRCh38
rs199508719 Chr.14: 90268459-90268459 on Build GRCh38
rs780335031 Chr.14: 90268438-90268438 on Build GRCh38
rs147061412 Chr.14: 90268375-90268375 on Build GRCh38
rs1021362234 Chr.14: 90268498-90268498 on Build GRCh38
rs772214892 Chr.14: 90268457-90268457 on Build GRCh38
rs61735655 Chr.14: 90268369-90268369 on Build GRCh38
rs986319722 Chr.14: 90268440-90268440 on Build GRCh38
rs373464261 Chr.14: 90268383-90268383 on Build GRCh38
rs767167551 Chr.14: 90268427-90268427 on Build GRCh38
rs376453812 Chr.14: 90268425-90268425 on Build GRCh38
rs148097391 Chr.14: 90268397-90268397 on Build GRCh38
rs1002277199 Chr.14: 90268505-90268505 on Build GRCh38
rs762847784 Chr.14: 90268396-90268396 on Build GRCh38
rs367968578 Chr.14: 90268495-90268495 on Build GRCh38
rs764345395 Chr.14: 90268485-90268485 on Build GRCh38
rs748640589 Chr.14: 90268465-90268469 on Build GRCh38
rs200738856 Chr.14: 90268458-90268458 on Build GRCh38
rs754204663 Chr.14: 90268447-90268447 on Build GRCh38
rs755472744 Chr.14: 90268452-90268452 on Build GRCh38
rs755783747 Chr.14: 90268433-90268433 on Build GRCh38
rs779517549 Chr.14: 90268453-90268453 on Build GRCh38
rs752396965 Chr.14: 90268481-90268481 on Build GRCh38
rs781710905 Chr.14: 90268433-90268433 on Build GRCh38
rs375519977 Chr.14: 90268462-90268462 on Build GRCh38
rs765045067 Chr.14: 90268445-90268445 on Build GRCh38
rs749973559 Chr.14: 90268429-90268429 on Build GRCh38
rs566417995 Chr.14: 90268496-90268496 on Build GRCh38
rs11624246 Chr.14: 90268482-90268482 on Build GRCh38
rs748253268 Chr.14: 90268456-90268456 on Build GRCh38
rs955657564 Chr.14: 90268488-90268488 on Build GRCh38
rs886440982 Chr.14: 90268350-90268350 on Build GRCh38

View Full Product Details