Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 28867240-28867511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH2B1
Entrez Gene ID
25970
Gene Name
SH2B adaptor protein 1
Gene Aliases
PSM, SH2B
UniGene
Hs.15744
Gene Chromosome Location
Chr.16: 28846600-28874213 on Build GRCh38
Transcripts
NM_001145795.1 NM_001145796.1 NM_001145797.1 NM_001145812.1 NM_001308293.1 NM_001308294.1 NM_015503.2 XM_017023114.1 XM_017023115.1 XM_017023116.1 XM_017023117.1 XM_017023118.1 XM_017023119.1 XM_017023120.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
471599 c.1023G>T p.E341D Chr.16: 28867414-28867414 on Build GRCh38
6001992 c.1015G>A p.D339N Chr.16: 28867406-28867406 on Build GRCh38
4725524 c.1040A>G p.K347R Chr.16: 28867431-28867431 on Build GRCh38
6452086 c.1029G>A p.T343T Chr.16: 28867420-28867420 on Build GRCh38
1518835 c.1001C>A p.A334D Chr.16: 28867392-28867392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375732131 Chr.16: 28867322-28867322 on Build GRCh38
rs781056810 Chr.16: 28867410-28867410 on Build GRCh38
rs763243760 Chr.16: 28867296-28867296 on Build GRCh38
rs774437479 Chr.16: 28867313-28867313 on Build GRCh38
rs1056899285 Chr.16: 28867457-28867457 on Build GRCh38
rs867936766 Chr.16: 28867341-28867341 on Build GRCh38
rs375143445 Chr.16: 28867315-28867315 on Build GRCh38
rs372521485 Chr.16: 28867427-28867427 on Build GRCh38
rs771083382 Chr.16: 28867298-28867298 on Build GRCh38
rs181530285 Chr.16: 28867280-28867280 on Build GRCh38
rs777071835 Chr.16: 28867475-28867475 on Build GRCh38
rs572568427 Chr.16: 28867490-28867490 on Build GRCh38
rs991528706 Chr.16: 28867295-28867295 on Build GRCh38
rs780187395 Chr.16: 28867384-28867384 on Build GRCh38
rs139298340 Chr.16: 28867419-28867419 on Build GRCh38
rs552659326 Chr.16: 28867465-28867465 on Build GRCh38
rs759576352 Chr.16: 28867314-28867314 on Build GRCh38
rs768412345 Chr.16: 28867409-28867409 on Build GRCh38
rs756979905 Chr.16: 28867338-28867338 on Build GRCh38
rs757950448 Chr.16: 28867380-28867380 on Build GRCh38
rs757127519 Chr.16: 28867420-28867420 on Build GRCh38
rs369196090 Chr.16: 28867379-28867379 on Build GRCh38
rs200316520 Chr.16: 28867458-28867458 on Build GRCh38
rs369977270 Chr.16: 28867321-28867321 on Build GRCh38
rs754082502 Chr.16: 28867337-28867337 on Build GRCh38
rs1027785641 Chr.16: 28867283-28867284 on Build GRCh38
rs186911218 Chr.16: 28867491-28867491 on Build GRCh38
rs767273639 Chr.16: 28867478-28867478 on Build GRCh38
rs935907949 Chr.16: 28867355-28867355 on Build GRCh38
rs947055242 Chr.16: 28867424-28867424 on Build GRCh38
rs759666297 Chr.16: 28867444-28867444 on Build GRCh38
rs576962727 Chr.16: 28867403-28867403 on Build GRCh38
rs537334816 Chr.16: 28867285-28867285 on Build GRCh38
rs764512179 Chr.16: 28867330-28867330 on Build GRCh38
rs751333768 Chr.16: 28867404-28867404 on Build GRCh38
rs765020900 Chr.16: 28867483-28867483 on Build GRCh38
rs958897311 Chr.16: 28867288-28867288 on Build GRCh38
rs556914868 Chr.16: 28867367-28867367 on Build GRCh38
rs772127866 Chr.16: 28867455-28867455 on Build GRCh38
rs902953864 Chr.16: 28867454-28867454 on Build GRCh38

View Full Product Details