Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 4856765-4857038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC23A2
Entrez Gene ID
9962
Gene Name
solute carrier family 23 member 2
Gene Aliases
NBTL1, SLC23A1, SVCT2, YSPL2, hSVCT2
UniGene
Hs.516866
Gene Chromosome Location
Chr.20: 4852356-5010293 on Build GRCh38
Transcripts
NM_005116.5 NM_203327.1 XM_011529414.2 XM_011529415.2 XM_011529416.2 XM_011529417.2 XM_017028171.1 XM_017028172.1 XM_017028173.1 XM_017028174.1 XM_017028175.1 XM_017028176.1 XM_017028177.1 XM_017028178.1 XM_017028179.1 XM_017028180.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1190206 c.1946C>T p.T649M Chr.20: 4856979-4856979 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936803226 Chr.20: 4856968-4856968 on Build GRCh38
rs371564157 Chr.20: 4856945-4856945 on Build GRCh38
rs72552229 Chr.20: 4856839-4856839 on Build GRCh38
rs1051400802 Chr.20: 4856857-4856857 on Build GRCh38
rs759370167 Chr.20: 4857006-4857006 on Build GRCh38
rs202082837 Chr.20: 4857014-4857014 on Build GRCh38
rs779381594 Chr.20: 4856951-4856951 on Build GRCh38
rs1034600080 Chr.20: 4856954-4856954 on Build GRCh38
rs143357840 Chr.20: 4856876-4856876 on Build GRCh38
rs755359657 Chr.20: 4856947-4856947 on Build GRCh38
rs918057376 Chr.20: 4856934-4856934 on Build GRCh38
rs958615643 Chr.20: 4856957-4856957 on Build GRCh38
rs200105638 Chr.20: 4857017-4857017 on Build GRCh38
rs770548154 Chr.20: 4856984-4856984 on Build GRCh38
rs770137235 Chr.20: 4856937-4856937 on Build GRCh38
rs147205702 Chr.20: 4856978-4856978 on Build GRCh38
rs1041802641 Chr.20: 4856795-4856795 on Build GRCh38
rs776126491 Chr.20: 4857015-4857016 on Build GRCh38
rs181330076 Chr.20: 4856981-4856981 on Build GRCh38
rs750524569 Chr.20: 4857018-4857018 on Build GRCh38
rs748686355 Chr.20: 4856956-4856956 on Build GRCh38
rs754878558 Chr.20: 4856979-4856979 on Build GRCh38
rs963159500 Chr.20: 4856943-4856943 on Build GRCh38
rs757323305 Chr.20: 4856816-4856816 on Build GRCh38
rs148350851 Chr.20: 4856913-4856913 on Build GRCh38
rs998844794 Chr.20: 4856841-4856841 on Build GRCh38
rs7343820 Chr.20: 4856880-4856880 on Build GRCh38
rs145230333 Chr.20: 4856799-4856799 on Build GRCh38
rs776326380 Chr.20: 4856986-4856986 on Build GRCh38
rs72552230 Chr.20: 4856885-4856885 on Build GRCh38
rs945910337 Chr.20: 4856987-4856987 on Build GRCh38
rs565897375 Chr.20: 4856794-4856794 on Build GRCh38
rs765051969 Chr.20: 4857013-4857013 on Build GRCh38
rs749660102 Chr.20: 4856946-4856946 on Build GRCh38
rs555195494 Chr.20: 4856803-4856803 on Build GRCh38
rs936158471 Chr.20: 4856804-4856804 on Build GRCh38
rs1004577336 Chr.20: 4856877-4856877 on Build GRCh38
rs762894105 Chr.20: 4857016-4857016 on Build GRCh38
rs537039640 Chr.20: 4856864-4856864 on Build GRCh38

View Full Product Details