Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80484464-80484689 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRQ
Entrez Gene ID
374462
Gene Name
protein tyrosine phosphatase, receptor type Q
Gene Aliases
DFNB84, DFNB84A, PTPGMC1, R-PTP-Q
UniGene
Hs.539284
Gene Chromosome Location
Chr.12: 80424261-80680273 on Build GRCh38
Transcripts
NM_001145026.1 XM_017019273.1 XM_017019274.1 XM_017019275.1

SNP Detail

SNP ID Chromosome Location
rs767695379 Chr.12: 80484548-80484548 on Build GRCh38
rs768790872 Chr.12: 80484613-80484614 on Build GRCh38
rs576490030 Chr.12: 80484634-80484634 on Build GRCh38
rs931970398 Chr.12: 80484565-80484565 on Build GRCh38
rs771983585 Chr.12: 80484643-80484643 on Build GRCh38
rs773869003 Chr.12: 80484587-80484587 on Build GRCh38
rs1023684712 Chr.12: 80484598-80484598 on Build GRCh38
rs1005570619 Chr.12: 80484537-80484537 on Build GRCh38
rs747612931 Chr.12: 80484592-80484592 on Build GRCh38
rs994932624 Chr.12: 80484490-80484490 on Build GRCh38
rs961025926 Chr.12: 80484582-80484582 on Build GRCh38
rs61729287 Chr.12: 80484531-80484531 on Build GRCh38
rs866766784 Chr.12: 80484629-80484629 on Build GRCh38
rs1041881258 Chr.12: 80484638-80484638 on Build GRCh38
rs987148620 Chr.12: 80484640-80484640 on Build GRCh38
rs554930585 Chr.12: 80484624-80484624 on Build GRCh38
rs968132035 Chr.12: 80484606-80484606 on Build GRCh38
rs907389130 Chr.12: 80484657-80484657 on Build GRCh38
rs779784942 Chr.12: 80484639-80484639 on Build GRCh38
rs778061943 Chr.12: 80484491-80484492 on Build GRCh38
rs577783320 Chr.12: 80484520-80484520 on Build GRCh38
rs190098322 Chr.12: 80484500-80484500 on Build GRCh38
rs747783884 Chr.12: 80484489-80484489 on Build GRCh38
rs377572771 Chr.12: 80484607-80484607 on Build GRCh38
rs773429164 Chr.12: 80484494-80484494 on Build GRCh38
rs766757071 Chr.12: 80484511-80484511 on Build GRCh38
rs753038688 Chr.12: 80484584-80484584 on Build GRCh38
rs57971665 Chr.12: 80484538-80484538 on Build GRCh38
rs1054502852 Chr.12: 80484571-80484571 on Build GRCh38
rs750132916 Chr.12: 80484628-80484628 on Build GRCh38
rs1049232863 Chr.12: 80484510-80484510 on Build GRCh38
rs372238123 Chr.12: 80484493-80484493 on Build GRCh38
rs534550525 Chr.12: 80484626-80484626 on Build GRCh38

View Full Product Details