Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41486193-41486454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT36
Entrez Gene ID
8689
Gene Name
keratin 36
Gene Aliases
HA6, KRTHA6, hHa6
UniGene
Hs.248189
Gene Chromosome Location
Chr.17: 41486136-41489864 on Build GRCh38
Transcripts
NM_003771.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4266875 c.1370C>T p.S457F Chr.17: 41486410-41486410 on Build GRCh38
5984154 c.1342G>A p.E448K Chr.17: 41486438-41486438 on Build GRCh38
6384158 c.1403G>T p.*468L Chr.17: 41486377-41486377 on Build GRCh38
3517402 c.1378G>A p.E460K Chr.17: 41486402-41486402 on Build GRCh38
7096367 c.1352G>A p.R451K Chr.17: 41486428-41486428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771669637 Chr.17: 41486386-41486386 on Build GRCh38
rs747417383 Chr.17: 41486328-41486329 on Build GRCh38
rs754790206 Chr.17: 41486284-41486284 on Build GRCh38
rs774104805 Chr.17: 41486328-41486330 on Build GRCh38
rs150827736 Chr.17: 41486419-41486419 on Build GRCh38
rs567416541 Chr.17: 41486223-41486223 on Build GRCh38
rs555765804 Chr.17: 41486396-41486396 on Build GRCh38
rs750760384 Chr.17: 41486344-41486344 on Build GRCh38
rs750793210 Chr.17: 41486425-41486425 on Build GRCh38
rs757498359 Chr.17: 41486325-41486325 on Build GRCh38
rs968845868 Chr.17: 41486275-41486275 on Build GRCh38
rs375069450 Chr.17: 41486421-41486421 on Build GRCh38
rs991532250 Chr.17: 41486239-41486239 on Build GRCh38
rs764284774 Chr.17: 41486408-41486408 on Build GRCh38
rs772901640 Chr.17: 41486387-41486387 on Build GRCh38
rs753646835 Chr.17: 41486269-41486269 on Build GRCh38
rs565828637 Chr.17: 41486406-41486408 on Build GRCh38
rs370656533 Chr.17: 41486376-41486376 on Build GRCh38
rs766176687 Chr.17: 41486224-41486224 on Build GRCh38
rs977700283 Chr.17: 41486290-41486290 on Build GRCh38
rs768000309 Chr.17: 41486339-41486339 on Build GRCh38
rs1041881170 Chr.17: 41486253-41486253 on Build GRCh38
rs754469844 Chr.17: 41486381-41486381 on Build GRCh38
rs886996592 Chr.17: 41486374-41486374 on Build GRCh38
rs372036536 Chr.17: 41486400-41486400 on Build GRCh38
rs1024743472 Chr.17: 41486395-41486395 on Build GRCh38
rs1004028918 Chr.17: 41486393-41486393 on Build GRCh38
rs142255355 Chr.17: 41486397-41486397 on Build GRCh38
rs924841018 Chr.17: 41486291-41486291 on Build GRCh38
rs537830816 Chr.17: 41486273-41486273 on Build GRCh38
rs745504438 Chr.17: 41486302-41486302 on Build GRCh38
rs774507782 Chr.17: 41486409-41486409 on Build GRCh38
rs148487920 Chr.17: 41486383-41486383 on Build GRCh38
rs2071602 Chr.17: 41486309-41486309 on Build GRCh38
rs897728831 Chr.17: 41486412-41486412 on Build GRCh38
rs762157702 Chr.17: 41486413-41486413 on Build GRCh38
rs946216386 Chr.17: 41486246-41486246 on Build GRCh38
rs944102200 Chr.17: 41486300-41486300 on Build GRCh38
rs940943576 Chr.17: 41486294-41486319 on Build GRCh38
rs914763707 Chr.17: 41486268-41486268 on Build GRCh38
rs190109383 Chr.17: 41486301-41486301 on Build GRCh38
rs185915535 Chr.17: 41486293-41486293 on Build GRCh38
rs914789749 Chr.17: 41486236-41486236 on Build GRCh38
rs554181711 Chr.17: 41486362-41486362 on Build GRCh38
rs756627658 Chr.17: 41486354-41486354 on Build GRCh38

View Full Product Details