Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155941942-155942189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXFP4
Entrez Gene ID
339403
Gene Name
relaxin/insulin like family peptide receptor 4
Gene Aliases
GPCR142, GPR100, RLN3R2, RXFPR4
UniGene
Hs.449914
Gene Chromosome Location
Chr.1: 155941689-155942834 on Build GRCh38
Transcripts
NM_181885.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461117 c.409G>A p.V137I Chr.1: 155942118-155942118 on Build GRCh38
3475084 c.436G>A p.A146T Chr.1: 155942145-155942145 on Build GRCh38
5511577 c.440C>T p.A147V Chr.1: 155942149-155942149 on Build GRCh38
2211358 c.402G>A p.A134A Chr.1: 155942111-155942111 on Build GRCh38
2211357 c.328G>A p.G110R Chr.1: 155942037-155942037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768269224 Chr.1: 155942054-155942054 on Build GRCh38
rs757610256 Chr.1: 155942093-155942093 on Build GRCh38
rs779193216 Chr.1: 155942131-155942131 on Build GRCh38
rs149283693 Chr.1: 155942117-155942117 on Build GRCh38
rs754256552 Chr.1: 155942025-155942025 on Build GRCh38
rs766529074 Chr.1: 155941998-155941998 on Build GRCh38
rs779687290 Chr.1: 155942121-155942121 on Build GRCh38
rs748840436 Chr.1: 155941964-155941964 on Build GRCh38
rs139318467 Chr.1: 155941967-155941967 on Build GRCh38
rs759675069 Chr.1: 155942081-155942081 on Build GRCh38
rs764348848 Chr.1: 155942169-155942169 on Build GRCh38
rs1042988959 Chr.1: 155942030-155942030 on Build GRCh38
rs958648993 Chr.1: 155942144-155942144 on Build GRCh38
rs746835195 Chr.1: 155942052-155942052 on Build GRCh38
rs748089131 Chr.1: 155942150-155942150 on Build GRCh38
rs577473640 Chr.1: 155942111-155942111 on Build GRCh38
rs144073711 Chr.1: 155942149-155942149 on Build GRCh38
rs753169719 Chr.1: 155942011-155942011 on Build GRCh38
rs764314560 Chr.1: 155942110-155942110 on Build GRCh38
rs200353483 Chr.1: 155942136-155942136 on Build GRCh38
rs150519113 Chr.1: 155942095-155942095 on Build GRCh38
rs201370533 Chr.1: 155942124-155942124 on Build GRCh38
rs754825959 Chr.1: 155942125-155942125 on Build GRCh38
rs761607398 Chr.1: 155942057-155942057 on Build GRCh38
rs199683371 Chr.1: 155942037-155942037 on Build GRCh38
rs778128008 Chr.1: 155941961-155941961 on Build GRCh38
rs548554375 Chr.1: 155942082-155942082 on Build GRCh38
rs371453354 Chr.1: 155942007-155942007 on Build GRCh38
rs768700062 Chr.1: 155941992-155941992 on Build GRCh38
rs778446357 Chr.1: 155942026-155942026 on Build GRCh38
rs757804806 Chr.1: 155942114-155942114 on Build GRCh38
rs760852516 Chr.1: 155942165-155942165 on Build GRCh38
rs375481936 Chr.1: 155942065-155942065 on Build GRCh38
rs746649818 Chr.1: 155942165-155942167 on Build GRCh38
rs771694127 Chr.1: 155941982-155941982 on Build GRCh38
rs200326480 Chr.1: 155942036-155942036 on Build GRCh38
rs887240205 Chr.1: 155941997-155941997 on Build GRCh38
rs770276231 Chr.1: 155941966-155941966 on Build GRCh38
rs895170850 Chr.1: 155942075-155942075 on Build GRCh38
rs773225872 Chr.1: 155942147-155942147 on Build GRCh38
rs775306235 Chr.1: 155941985-155941985 on Build GRCh38
rs751854013 Chr.1: 155942006-155942006 on Build GRCh38
rs200703792 Chr.1: 155942142-155942142 on Build GRCh38
rs757849833 Chr.1: 155942118-155942118 on Build GRCh38
rs756372124 Chr.1: 155942012-155942012 on Build GRCh38
rs764610139 Chr.1: 155941981-155941982 on Build GRCh38
rs745946606 Chr.1: 155942148-155942148 on Build GRCh38
rs931520740 Chr.1: 155941983-155941983 on Build GRCh38
rs760387289 Chr.1: 155941991-155941991 on Build GRCh38
rs754332239 Chr.1: 155942071-155942072 on Build GRCh38
rs778343223 Chr.1: 155942024-155942024 on Build GRCh38
rs369923925 Chr.1: 155942066-155942066 on Build GRCh38
rs779671231 Chr.1: 155942041-155942041 on Build GRCh38
rs747969629 Chr.1: 155942137-155942137 on Build GRCh38
rs367884988 Chr.1: 155942017-155942017 on Build GRCh38
rs775505351 Chr.1: 155942090-155942090 on Build GRCh38
rs944131513 Chr.1: 155942005-155942005 on Build GRCh38
rs745471725 Chr.1: 155941979-155941979 on Build GRCh38
rs776375640 Chr.1: 155941994-155941994 on Build GRCh38
rs769809373 Chr.1: 155942140-155942140 on Build GRCh38
rs146210789 Chr.1: 155942127-155942127 on Build GRCh38
rs767845245 Chr.1: 155942009-155942009 on Build GRCh38
rs776663883 Chr.1: 155942056-155942056 on Build GRCh38

View Full Product Details