Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76376089-76376350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
UniGene
Hs.458986
Gene Chromosome Location
Chr.15: 76348185-76905455 on Build GRCh38
Transcripts
NM_001145923.1 NM_020843.2 XM_005254417.2 XM_005254419.2 XM_011521650.1 XM_011521652.2 XM_011521653.2 XM_011521654.2 XM_011521656.2 XM_017022266.1 XM_017022267.1 XM_017022268.1 XM_017022269.1 XM_017022270.1 XM_017022271.1 XM_017022272.1 XM_017022273.1 XM_017022275.1 XM_017022276.1 XM_017022277.1 XM_017022278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7388750 c.3028G>T p.G1010C Chr.15: 76376251-76376251 on Build GRCh38
3504179 c.3845C>A p.P1282Q Chr.15: 76376172-76376172 on Build GRCh38
3504178 c.3107C>A p.P1036Q Chr.15: 76376172-76376172 on Build GRCh38
1608704 c.2981C>T p.A994V Chr.15: 76376298-76376298 on Build GRCh38
3504181 c.3758C>T p.S1253F Chr.15: 76376259-76376259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374019205 Chr.15: 76376237-76376237 on Build GRCh38
rs754930859 Chr.15: 76376246-76376246 on Build GRCh38
rs1008469377 Chr.15: 76376313-76376313 on Build GRCh38
rs768530315 Chr.15: 76376259-76376259 on Build GRCh38
rs527875423 Chr.15: 76376272-76376272 on Build GRCh38
rs762610169 Chr.15: 76376250-76376250 on Build GRCh38
rs757336864 Chr.15: 76376145-76376145 on Build GRCh38
rs981794229 Chr.15: 76376139-76376139 on Build GRCh38
rs761222341 Chr.15: 76376307-76376307 on Build GRCh38
rs746670301 Chr.15: 76376203-76376203 on Build GRCh38
rs889483980 Chr.15: 76376269-76376269 on Build GRCh38
rs750570976 Chr.15: 76376288-76376288 on Build GRCh38
rs767697314 Chr.15: 76376275-76376275 on Build GRCh38
rs934469787 Chr.15: 76376147-76376147 on Build GRCh38
rs766744951 Chr.15: 76376308-76376308 on Build GRCh38
rs567097821 Chr.15: 76376263-76376263 on Build GRCh38
rs373336878 Chr.15: 76376163-76376163 on Build GRCh38
rs199940124 Chr.15: 76376206-76376206 on Build GRCh38
rs972307725 Chr.15: 76376291-76376291 on Build GRCh38
rs529438073 Chr.15: 76376125-76376125 on Build GRCh38
rs775337330 Chr.15: 76376247-76376247 on Build GRCh38
rs549199597 Chr.15: 76376330-76376330 on Build GRCh38
rs755232490 Chr.15: 76376328-76376328 on Build GRCh38
rs374119195 Chr.15: 76376144-76376144 on Build GRCh38
rs774654951 Chr.15: 76376270-76376270 on Build GRCh38
rs770371959 Chr.15: 76376204-76376204 on Build GRCh38
rs750948803 Chr.15: 76376151-76376151 on Build GRCh38
rs201658000 Chr.15: 76376173-76376173 on Build GRCh38
rs374150740 Chr.15: 76376331-76376331 on Build GRCh38
rs745802052 Chr.15: 76376228-76376228 on Build GRCh38
rs374983073 Chr.15: 76376178-76376178 on Build GRCh38
rs903322022 Chr.15: 76376151-76376151 on Build GRCh38
rs947613199 Chr.15: 76376134-76376134 on Build GRCh38
rs141924677 Chr.15: 76376131-76376131 on Build GRCh38
rs780435349 Chr.15: 76376171-76376171 on Build GRCh38
rs777250353 Chr.15: 76376195-76376195 on Build GRCh38
rs773852040 Chr.15: 76376153-76376153 on Build GRCh38
rs754399001 Chr.15: 76376326-76376326 on Build GRCh38
rs377734546 Chr.15: 76376149-76376149 on Build GRCh38
rs1035555157 Chr.15: 76376301-76376301 on Build GRCh38
rs371119046 Chr.15: 76376154-76376154 on Build GRCh38
rs763208532 Chr.15: 76376128-76376128 on Build GRCh38

View Full Product Details