Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 960322-960587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPO4
Entrez Gene ID
343637
Gene Name
R-spondin 4
Gene Aliases
C20orf182, CRISTIN4
UniGene
Hs.444980
Gene Chromosome Location
Chr.20: 958452-1002264 on Build GRCh38
Transcripts
NM_001029871.3 NM_001040007.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6029396 c.635G>A p.R212H Chr.20: 960427-960427 on Build GRCh38
5392682 c.681G>A p.P227P Chr.20: 960381-960381 on Build GRCh38
4137183 c.697C>A p.Q233K Chr.20: 960365-960365 on Build GRCh38
4389240 c.672C>T p.D224D Chr.20: 960390-960390 on Build GRCh38
4980273 c.680C>T p.P227L Chr.20: 960382-960382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374615663 Chr.20: 960489-960489 on Build GRCh38
rs941829816 Chr.20: 960352-960352 on Build GRCh38
rs202178299 Chr.20: 960428-960428 on Build GRCh38
rs774771039 Chr.20: 960396-960396 on Build GRCh38
rs760903291 Chr.20: 960411-960411 on Build GRCh38
rs1045352878 Chr.20: 960554-960554 on Build GRCh38
rs766487997 Chr.20: 960343-960343 on Build GRCh38
rs369323347 Chr.20: 960506-960506 on Build GRCh38
rs767423520 Chr.20: 960431-960431 on Build GRCh38
rs963091399 Chr.20: 960446-960446 on Build GRCh38
rs375728608 Chr.20: 960373-960373 on Build GRCh38
rs768182696 Chr.20: 960389-960389 on Build GRCh38
rs773501751 Chr.20: 960410-960410 on Build GRCh38
rs201162335 Chr.20: 960382-960382 on Build GRCh38
rs576270623 Chr.20: 960476-960476 on Build GRCh38
rs374130199 Chr.20: 960424-960424 on Build GRCh38
rs930180877 Chr.20: 960353-960353 on Build GRCh38
rs188281610 Chr.20: 960482-960482 on Build GRCh38
rs973249616 Chr.20: 960354-960354 on Build GRCh38
rs369687640 Chr.20: 960422-960422 on Build GRCh38
rs755822660 Chr.20: 960455-960455 on Build GRCh38
rs1009410860 Chr.20: 960409-960409 on Build GRCh38
rs983007174 Chr.20: 960508-960508 on Build GRCh38
rs377200892 Chr.20: 960421-960421 on Build GRCh38
rs937989961 Chr.20: 960528-960528 on Build GRCh38
rs915481834 Chr.20: 960548-960548 on Build GRCh38
rs757255412 Chr.20: 960430-960430 on Build GRCh38
rs950620900 Chr.20: 960486-960486 on Build GRCh38
rs1042605994 Chr.20: 960402-960402 on Build GRCh38
rs949643224 Chr.20: 960532-960532 on Build GRCh38
rs774244770 Chr.20: 960542-960542 on Build GRCh38
rs918153061 Chr.20: 960526-960526 on Build GRCh38
rs11350888 Chr.20: 960540-960540 on Build GRCh38
rs760773975 Chr.20: 960559-960559 on Build GRCh38
rs951599368 Chr.20: 960491-960491 on Build GRCh38
rs902915706 Chr.20: 960406-960406 on Build GRCh38
rs762295741 Chr.20: 960400-960400 on Build GRCh38
rs765335066 Chr.20: 960427-960427 on Build GRCh38
rs563843176 Chr.20: 960371-960371 on Build GRCh38
rs745580942 Chr.20: 960380-960380 on Build GRCh38
rs113093200 Chr.20: 960355-960355 on Build GRCh38
rs781119266 Chr.20: 960379-960379 on Build GRCh38
rs759063553 Chr.20: 960390-960390 on Build GRCh38
rs866145059 Chr.20: 960456-960456 on Build GRCh38
rs772494608 Chr.20: 960401-960401 on Build GRCh38
rs751507935 Chr.20: 960372-960372 on Build GRCh38
rs750184862 Chr.20: 960442-960442 on Build GRCh38
rs534275132 Chr.20: 960347-960347 on Build GRCh38
rs369860447 Chr.20: 960381-960381 on Build GRCh38

View Full Product Details